Skip to Content
Merck
All Photos(1)

Key Documents

15-2230

Sigma-Aldrich

2-Methylbutane

SAJ first grade, ≥99.0%

Synonym(s):

Isopentane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(CH3)2
CAS Number:
Molecular Weight:
72.15
Beilstein:
1730723
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:

grade

SAJ first grade

vapor density

2.6 (vs air)

vapor pressure

11.17 psi ( 20 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

788 °F

expl. lim.

8.3 %

availability

available only in Japan

refractive index

n20/D 1.354 (lit.)

bp

30 °C (lit.)

mp

-160 °C (lit.)

density

0.62 g/mL at 25 °C (lit.)

SMILES string

CCC(C)C

InChI

1S/C5H12/c1-4-5(2)3/h5H,4H2,1-3H3

InChI key

QWTDNUCVQCZILF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 1 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-59.8 °F - closed cup

Flash Point(C)

-51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicole Antonio et al.
The EMBO journal, 34(17), 2219-2236 (2015-07-03)
There is a long-standing association between wound healing and cancer, with cancer often described as a "wound that does not heal". However, little is known about how wounding, such as following surgery, biopsy collection or ulceration, might impact on cancer
Susanne Steu et al.
Virchows Archiv : an international journal of pathology, 452(3), 305-312 (2008-02-07)
Different methods for snap freezing surgical human tissue specimens exist. At pathology institutes with higher work loads, solid carbon dioxide, freezing sprays, and cryostat freezing are commonly used as coolants for diagnosing frozen tissue sections, whereas for tissue banking, liquid
Kariem Ezzat et al.
Nano letters, 15(7), 4364-4373 (2015-06-05)
Antisense oligonucleotides (ASOs) have the potential to revolutionize medicine due to their ability to manipulate gene function for therapeutic purposes. ASOs are chemically modified and/or incorporated within nanoparticles to enhance their stability and cellular uptake, however, a major challenge is
Emma Sandén et al.
Journal of neuro-oncology, 123(1), 1-13 (2015-03-31)
The CD24 glycoprotein is a mediator of neuronal proliferation, differentiation and immune suppression in the normal CNS, and a proposed cancer biomarker in multiple peripheral tumor types. We performed a comparative analysis of CD24 gene expression in a large cohort
Maarten Ooms et al.
Nuclear medicine and biology, 41(8), 695-704 (2014-07-09)
In vivo visualization of PDE10A using PET provides a tool to evaluate the role of PDE10A in various neuropsychiatric diseases and can also be useful in the clinical evaluation of PDE10A inhibitor drug candidates. We evaluated several carbon-11 and fluorine-18

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service