Skip to Content
Merck
All Photos(1)

Key Documents

15-0800

Sigma-Aldrich

Iodoform

SAJ first grade

Synonym(s):

Triiodomethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHI3
CAS Number:
Molecular Weight:
393.73
Beilstein:
1697010
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
grade:
SAJ first grade

grade

SAJ first grade

Assay

≥99.0%

form

solid

availability

available only in Japan

dilution

(for analytical testing)

mp

118-121 °C (lit.)

density

4.008 g/mL at 25 °C (lit.)

SMILES string

IC(I)I

InChI

1S/CHI3/c2-1(3)4/h1H

InChI key

OKJPEAGHQZHRQV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hervé Gallard et al.
Environmental science & technology, 43(18), 7003-7009 (2009-10-08)
This study shows that iodinated organic compounds can be produced when iodide-containing waters are in contact with manganese oxide birnessite (delta-MnO2) in the pH range of 5-7. In the absence of natural organic matter (NOM), iodide is oxidized to iodate
Eduardo Antunes Bortoluzzi et al.
Oral surgery, oral medicine, oral pathology, oral radiology, and endodontics, 108(4), 628-632 (2009-08-25)
The objective of this study was to evaluate the radiopacity of white Portland cement (PC) associated with bismuth oxide (Bi(2)O(3)), barium sulfate (BaSO(4)), iodoform (CHI(3)), and zirconium oxide (ZrO(2)). White mineral trioxide aggregate (WMTA) and PC without radiopacifier were used
Raimunda Vilmar Evangelista Lima et al.
Pediatric dentistry, 33(1), 14-18 (2011-03-17)
This study's purpose was to histologically evaluate the repair of rat tissue after pulpotomy and covering the pulp tissue with Copaifera langsdorffi oil resin, green propolis extract, fibrin sponge and iodoform-based paste. Pulpotomies were performed in the maxillary and mandibular
Yi-Pin Lin et al.
Environmental science & technology, 42(8), 2919-2924 (2008-05-24)
Lead oxide (PbO2) can be an important form of lead mineral scale occurring in some water distribution systems. It is believed to be formed by the oxidation of lead-containing plumbing materials by free chlorine. Its reactivity in water, however, has
Moti Moskovitz et al.
Quintessence international (Berlin, Germany : 1985), 43(5), 361-368 (2012-04-27)
To compare the degree of root resorption in endodontically treated primary molars with that of homologous teeth without root canal treatment. A retrospective study was carried out comprising 105 records of children who had received root canal treatment in a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service