Skip to Content
Merck
All Photos(1)

Key Documents

12-0440

Sigma-Aldrich

Giemsa stain solution

Synonym(s):

Giemsa stain, Azure mixture sicc. Giemsa stain

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14ClN3S
CAS Number:
Molecular Weight:
291.80
MDL number:
UNSPSC Code:
41116100
PubChem Substance ID:

availability

available only in Japan

mp

300 °C (lit.)

SMILES string

[Cl-].CN(C)c1ccc2N=C3C=CC(=[NH2+])C=C3Sc2c1

InChI

1S/C14H13N3S.ClH/c1-17(2)10-4-6-12-14(8-10)18-13-7-9(15)3-5-11(13)16-12;/h3-8,15H,1-2H3;1H

InChI key

NALREUIWICQLPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Suitability

blood stain

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service