Skip to Content
Merck
All Photos(1)

Documents

11-0750

Sigma-Aldrich

Formamide

SAJ first grade, ≥98.5%

Synonym(s):

Amide C1, Formic amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCONH2
CAS Number:
Molecular Weight:
45.04
Beilstein:
505995
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:

grade

SAJ first grade

vapor density

1.55 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)
30 mmHg ( 129 °C)

Assay

≥98.5%

form

liquid

autoignition temp.

932 °F

expl. lim.

2.7-19 % (lit.)

availability

available only in Japan

technique(s)

hybridization: suitable

refractive index

n20/D 1.447 (lit.)

pH

4-10 (20 °C, 200 g/L)

bp

210 °C (lit.)

mp

2-3 °C (lit.)

density

1.134 g/mL at 25 °C (lit.)

SMILES string

NC=O

InChI

1S/CH3NO/c2-1-3/h1H,(H2,2,3)

InChI key

ZHNUHDYFZUAESO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Formamide destabilizes nucleic acid duplexes and may be used, typically, at a concentration of 50%, in hybridization protocols requiring lower hybridization temperatures.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

305.6 °F

Flash Point(C)

152 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Chen et al.
Prenatal diagnosis, 35(6), 534-543 (2015-02-03)
To develop a single-tube multi-marker assay for improved preimplantation genetic diagnosis (PGD) of deletional and/or non-deletional Hb Bart's hydrops fetalis syndrome, providing haplotype confirmation of deletional status, and maximization of linkage informativity. We performed in silico mining to identify novel
Mado Vandewoestyne et al.
International journal of legal medicine, 123(5), 441-447 (2009-03-26)
Laser microdissection is a valuable tool for isolating specific cells from mixtures, such as male cells in a mixture with female cells, e.g., in cases of sexual assault. These cells can be stained with Y-chromosome-specific probes. We developed an automatic
Penelope A Bryant et al.
PloS one, 6(5), e19556-e19556 (2011-06-10)
A central issue in the design of microarray-based analysis of global gene expression is that variability resulting from experimental processes may obscure changes resulting from the effect being investigated. This study quantified the variability in gene expression at each level
A Touati et al.
Journal of clinical microbiology, 53(10), 3182-3194 (2015-07-24)
Molecular typing of Mycoplasma pneumoniae is an important tool for identifying grouped cases and investigating outbreaks. In the present study, we developed a new genotyping method based on single nucleotide polymorphisms (SNPs) selected from the whole-genome sequencing of eight M.
Jing Wang et al.
The journal of physical chemistry. B, 117(8), 2314-2320 (2013-01-26)
A step-by-step mechanistic pathway following the transformation of formamide to purine through a five-membered ring intermediate has been explored by density functional theory computations. The highlight of the mechanistic route detailed here is that the proposed pathway represents the simplest

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service