Skip to Content
Merck
All Photos(1)

Key Documents

05-4580

Sigma-Aldrich

Chromium standard solution

suitable for atomic absorption spectrometry, 1 mg/mL Cr, 1000 ppm Cr

Synonym(s):

Potassium dichromate solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2Cr2O7
CAS Number:
Molecular Weight:
294.18
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

form

liquid

availability

available only in Japan

concentration

1 mg/mL Cr
1000 ppm Cr

suitability

suitable for atomic absorption spectrometry

SMILES string

[K+].[K+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O

InChI

1S/2Cr.2K.7O/q;;2*+1;;;;;;2*-1

InChI key

KMUONIBRACKNSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Muta. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neila Marouani et al.
Reproductive biology, 12(2), 119-133 (2012-08-02)
Hexavalent chromium is an environmental contaminant which may be associated with reproductive abnormalities in male rats. In the present study, we examined the effect of hexavalent chromium on male reproductive function of rats. Male Wistar rats received a daily intraperitoneal
Eduardo Molina-Jijón et al.
Toxicology, 291(1-3), 93-101 (2011-11-26)
Deferoxamine (DFO) is a recognized iron chelator which has been shown to exert nephroprotection in models of toxic nephropathies. In the present work the potential protective effects of DFO against Cr(VI)-induced nephrotoxicity and oxidant stress were evaluated. Rats were injected
C F C Araújo et al.
Ecotoxicology and environmental safety, 89, 212-221 (2013-01-22)
Microalgae are among the organisms that are most frequently recommended to assess water toxicity levels. They provide qualitative and quantitative population changes and have become important in environmental monitoring. The aim of the present study was to assess the sensitivity
Alena Machovcova
Acta dermatovenerologica Croatica : ADC, 20(2), 75-79 (2012-06-26)
The objective of our study was to determine the prevalence of contact hypersensitivity in asymptomatic Czech schoolchildren. We recruited 236 Prague asymptomatic schoolchildren for epicutaneous tests. The children were tested with European baseline series (25 allergens). A total of 218
Nejla Soudani et al.
Journal of physiology and biochemistry, 68(3), 397-409 (2012-02-22)
Chromium is known for its wide toxic manifestations. This experiment aims to evaluate the effect of selenium against oxidative stress induced by chromium in the cerebrum and cerebellum. Female Wistar rats were randomly divided into four groups of six each:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service