Skip to Content
Merck
All Photos(1)

Key Documents

03-4521

Sigma-Aldrich

Butyl acetate

≥99.5%, suitable for atomic absorption spectrometry

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COO(CH2)3CH3
CAS Number:
Molecular Weight:
116.16
Beilstein:
1741921
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

vapor density

4 (vs air)

vapor pressure

15 mmHg ( 25 °C)
8 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

790 °F

expl. lim.

7.6 %

availability

available only in Japan

refractive index

n20/D 1.394 (lit.)

pH

6.2 (20 °C, 5 g/L)

bp

124-126 °C (lit.)

mp

−78 °C (lit.)

density

0.88 g/mL at 25 °C (lit.)

suitability

suitable for atomic absorption spectrometry

SMILES string

CCCCOC(C)=O

InChI

1S/C6H12O2/c1-3-4-5-8-6(2)7/h3-5H2,1-2H3

InChI key

DKPFZGUDAPQIHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sami H Ali et al.
Bioresource technology, 102(21), 10094-10103 (2011-09-13)
Butyl acetate holds great potential as a sustainable biofuel additive. Heterogeneously catalyzed transesterification of biobutanol and bioethylacetate can produce butyl acetate. This route is eco-friendly and offers several advantages over the commonly used Fischer Esterification. The Amberlite IR 120- and
Yung-Chieh Tsao et al.
The Science of the total environment, 409(17), 3158-3165 (2011-05-31)
This study evaluated the efficacy of simultaneously employing three open-path Fourier transform infrared (OP-FTIR) spectrometers with 3-day consecutive monitoring, using an odor episode as an example. The corresponding monitoring paths were allocated among the possible emission sources of a semiconductor
Subhash Bhatia et al.
Journal of hazardous materials, 163(1), 73-81 (2008-07-25)
Adsorption behaviours of butyl acetate in air have been studied over silver-loaded Y (Si/Al=40) and ZSM-5 (Si/Al=140) zeolites. The silver metal was loaded into the zeolites by ion exchange (IE) and impregnation (IM) methods. The adsorption study was mainly conducted
Matias Rauma et al.
Journal of pharmaceutical sciences, 98(11), 4365-4375 (2009-03-14)
Recent work in our laboratory shows that it is possible to reproducibly measure the weight change over time of a piece of porcine skin exposed to a chemical vapor by thermogravimetric analysis (TGA), that is, using a microbalance maintained at
Prateek Kathel et al.
ISA transactions, 49(1), 130-137 (2009-10-10)
This work deals with the dynamics and control of a high-purity batch distillation column with chemical reaction. A heterogeneous esterification reaction between the acetic acid and butanol takes place to produce butyl acetate. The process model is formulated considering variable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service