Skip to Content
Merck
All Photos(1)

Key Documents

03-3734

Sigma-Aldrich

5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside

Synonym(s):

5-Bromo-4-chloro-3-indolyl β-D-galactoside, BCIG, X-Gal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15BrClNO6
CAS Number:
Molecular Weight:
408.63
Beilstein:
1402009
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

availability

available only in Japan

SMILES string

OC[C@H]1O[C@@H](Oc2c[nH]c3ccc(Br)c(Cl)c23)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C14H15BrClNO6/c15-5-1-2-6-9(10(5)16)7(3-17-6)22-14-13(21)12(20)11(19)8(4-18)23-14/h1-3,8,11-14,17-21H,4H2/t8-,11+,12+,13-,14-/m1/s1

InChI key

OPIFSICVWOWJMJ-AEOCFKNESA-N

Looking for similar products? Visit Product Comparison Guide

Application

X-Gal is a chromogenic substrate for β-galactosidase that produces a rich blue color that can easily be detected visually over background. X-Gal is the substrate of choice for blue-white selection of recombinant bacterial colonies with the lac+ genotype.

Suitability

for biological purposes

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yibing Wang et al.
PloS one, 8(3), e59195-e59195 (2013-03-26)
Our study had three objectives: to extend the plasmid-based transformation protocol to a clinical isolate of C. trachomatis belonging to the trachoma biovar, to provide "proof of principle" that it is possible to "knock out" selected plasmid genes (retaining a
Amy McCurley et al.
Nature medicine, 18(9), 1429-1433 (2012-08-28)
Hypertension is a cardiovascular risk factor present in over two-thirds of people over age 60 in North America; elevated blood pressure correlates with increased risk of heart attack, stroke and progression to heart and kidney failure. Current therapies are insufficient
Masahide Sakabe et al.
Development (Cambridge, England), 139(2), 385-395 (2011-12-22)
The progress of molecular genetics has enabled us to identify the genes responsible for congenital heart malformations. However, recent studies suggest that congenital heart diseases are induced not only by mutations in certain genes, but also by abnormal maternal factors.
Florian Weinberger et al.
Circulation research, 110(10), 1303-1310 (2012-03-20)
The transcription factor Islet-1 is a marker of cardiovascular progenitors during embryogenesis. The isolation of Islet-1-positive (Islet-1(+)) cells from early postnatal hearts suggested that Islet-1 also marks cardiac progenitors in adult life. We investigated the distribution and identity of Islet-1(+)
Enzo R Porrello et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(1), 187-192 (2012-12-19)
We recently identified a brief time period during postnatal development when the mammalian heart retains significant regenerative potential after amputation of the ventricular apex. However, one major unresolved question is whether the neonatal mouse heart can also regenerate in response

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service