Skip to Content
Merck

COLLB-RO

Roche

Collagenase B

from Clostridium histolyticum

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352204
EC Number:
NACRES:
NA.54
Biological source:
Clostridium histolyticum
Concentration:
0.5-2.5 mg/mL
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Clostridium histolyticum

sterility

non-sterile

form

lyophilized

packaging

pkg of 100 mg (11088807001), pkg of 2.5 g (11088831001), pkg of 500 mg (11088815001)

manufacturer/tradename

Roche

concentration

0.5-2.5 mg/mL

optimum pH

6.0-8.0

application(s)

sample preparation

storage temp.

2-8°C

General description

Collagenase B is prepared from Clostridium histolyticum cultures by filtration, ammonium sulfate precipitation, dialysis, and lyophilization. Collagenase from Clostridium histolyticum is a collagenolytic enzyme. It interacts with collagen-like substances and is responsible for the degradation of collagen into small peptides. It is widely used for the disaggregation of many types of tissues (e.g., lung, heart, muscle, bone, adipose tissue, liver, kidney, cartilage, mammary gland, placentae, blood vessels, brain, tumors) and for the preparation of single-cell suspensions for the establishment of primary cell culture systems. Collagenase B is recommended when yield and viability are important.

Application

Clostridium collagenase from Roche has been used to prepare cells from many types of tissue, such as hepatocytes, adipocytes, pancreatic islets, epithelial cells, muscle cells, endothelial cells, etc. However, suitability of each lot of the enzyme for disruption of a particular tissue should be determined empirically.
Collagenase B has been used for preparation of cells from mouse aortas and colorectal cancer tissue.

Biochem/physiol Actions

Collagenase degrades native collagen. Clostripain, trypsin-like enzymes, and neutral proteases degrade other proteins as well.

Features and Benefits

Enzyme activity:
Collagenase activity: >0.15U/mg (according to Wünsch) (+25°C, 4-phenyl-azobenzyl-oxycarbonyl-Pro-Leu-Gly-Pro-D-Arg as the substrate)
Contaminating enzyme activities: trypsin, clostripain, and total proteolytic activity
Collagenase B has a normal to high collagenase activity and a higher than average clostripain activity (usually >10U/mg).

Inhibitors:
Collagenase inhibitors: EDTA, EGTA, Cys, His, DTT, 2-mercaptoethanol
Collagenase is not inhibited by serum.
Clostripain inhibitors: TLCK
Trypsin inhibitors: aprotinin, trypsin inhibitor (egg white, soybean), serum

Preparation Note

Activator: Ca2+
Working concentration: 0.5 to 2.5 mg/ml
Storage conditions (working solution): -15 to -25 °C
Roche recommends reconstituting only the amount of lyophilizate needed for immediate use. The reconstituted solution can be stored at -15 to -25 °C for up to one week. Avoid repeated freezing and thawing since activity decreases after reconstitution.
Reconstitution in any balanced salt solution (e.g., HBSS)

Other Notes

Collagenase from Roche is assayed in Wünsch units (1 μmol of product formed per minute at +25 °C with Wünsch substrate).
Frequently, collagenase activities are given in Mandl units (1 μmol leucine liberated from collagen in 5 hours at +37 °C).
Unfortunately, there is no consistent conversion factor between the two units of activity, since the Mandl unit depends, in part, on the concentration of contaminating proteases in the collagenase preparation, an indefinable variable. A purer collagenase preparation would actually give a lower specific activity in Mandl units than a crude preparation. Clostridium preparations typically give conversion factors of approximately 1:1800 (e.g., a particular lot of Clostridium collagenase contained approximately 0.15 Wünsch U/mg and 250 Mandl U/mg).
For life science research only. Not for use in diagnostic procedures.


pictograms

Exclamation markHealth hazard

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Bo Jin et al.
Laboratory investigation; a journal of technical methods and pathology, 85(8), 992-1002 (2005-06-21)
The regression of cirrhosis is associated with increased intrahepatic collagenolytic enzyme activity. We investigated whether collagenase supplementation via portal vein infusion can retard cirrhosis development and/or reverse cirrhosis. In all, 35 rabbits were initially assigned to study. However, because of
Hongming Miao et al.
Nature communications, 7, 11716-11716 (2016-05-18)
Metabolic reprogramming in stromal cells plays an essential role in regulating tumour growth. The metabolic activities of tumour-associated macrophages (TAMs) in colorectal cancer (CRC) are incompletely characterized. Here, we identify TAM-derived factors and their roles in the development of CRC.
Wuzhou Wan et al.
Cardiovascular research, 97(3), 580-588 (2012-11-28)
Recent evidence suggests that both Ccr7 and its ligands, Ccl19 and Ccl21, are present in mouse and human atherosclerotic plaques; however, the role of Ccr7 in atherogenesis is still controversial. Here, we addressed this question by using the classic apolipoprotein



Global Trade Item Number

SKUGTIN
1108881500104061838250599
1108880700104061838698360
1108883100104061838250605