Skip to Content
Merck
All Photos(1)

Key Documents

11585878001

Roche

hGH ELISA

sufficient for 192 tests, >98% (SDS-PAGE), kit of 1, suitable for ELISA

Synonym(s):

ELISA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158

Assay

>98% (SDS-PAGE)

usage

sufficient for 192 tests

packaging

kit of 1

manufacturer/tradename

Roche

technique(s)

ELISA: suitable

storage temp.

2-8°C

General description

Human growth hormone (hGH) is specifically detected by the hGH ELISA. It does not cross-react with rat growth hormone. Cross-reactivity with the human homologues TSH, FSH, and LH has not been tested. The hGH protein from E. coli, included in the kit for the purpose of compiling a standard calibration curve, is provided with lot-specific content data as determined by immunoassay.

hGH Standards

The hGH protein from E. coli, included in the kit for the purpose of compiling a standard calibration curve, is provided with lot-specific content data as determined by immunoassay.
The hGH assay system differs from most of the other commonly used reporter proteins (CAT, β-Gal, or luciferase) in one important respect: hGH is a secreted protein, and is measured using samples of culture-medium supernatants thus (i) avoiding the necessity to lyse cells, (ii) allowing continuous monitoring of transient expression kinetics, and (iii) allowing use of the cells for other purposes (e.g., RNA isolation). Additionally, hGH is a mammalian gene that may contribute to the apparent high stability of the hGH mRNA in most mammalian cells.
Using the standard isotopic protocol, secretion of hGH protein into the culture-medium supernatant can be monitored approximately 24hours post-transfection, depending on the cell type and the sensitivity of the assay used. Usually, hGH is quantified by an immunoradiometric assay (IRMA), resulting in a linear range of approximately 0.1 to 50ng hGH/ml.

Specificity

The hGH ELISA specifically detecs human growth hormone (hGH). It does not cross-react with rat growth hormone.

Application

For research use only. Not for use in diagnostic procedures.
The hGH ELISA is used to quantitatively measure the expression level of hGH released into cell culture supernatant of eukaryotic cells transfected with a plasmid containing a reporter gene that encodes for hGH.

Features and Benefits

  • Standardized: Allows direct comparison of data from different sets of experiments, even when kits from different production lots are used
  • More sensitive: The hGH ELISA is approximately 20-times more sensitive than isotopic hGH assays
  • More specific: The hGH ELISA shows no crossreactivity with rat growth hormone
  • Faster: Measures hGH expression as early as 18hours post-transfection and requires only approximately 4 hours from start to finish
  • Easy to perform: Follows a standard ELISA protocol

Packaging

1 kit containing 9 components.

Specifications

Assay time: Approximately 4 hours
Sample material: Culture supernatant
Sensitivity: ≥5pg/ml (≥1pg/well)
Standards: The hGH protein from E. coli, included in the kit for the purpose of compiling a standard calibration curve, is provided with lot-specific content data as determined by immunoassay.

Principle

The hGH ELISA is based on the sandwich ELISA principle. Antibodies to hGH (anti-hGH) are prebound to the surface of the microplate. The culture supernatant, which contains secreted hGH, is added to the wells of the microplate modules. All hGH contained in the medium binds to the anti-hGH bound to the microplate surface. Next, a digoxigenin-labeled antibody to hGH (anti-hGH-DIG) is added, then bound to hGH. In the following step, an antibody to digoxigenin conjugated to peroxidase (anti-DIG-POD) is added, then bound to digoxigenin. In the final step, the peroxidase substrate ABTS is added. The peroxidase enzyme catalyzes the cleavage of the substrate, yielding a colored reaction product. The absorbance of the sample is determined using an ELISA reader, and is directly correlated to the level of hGH present in the medium supernatant. The sensitivity of the assay can be enhanced using the peroxidase substrate (ABTS) with substrate enhancer.

Preparation Note

Working solution: Solution 1 hGH Stock Solution (bottle 1)
Reconstitute the lyophilizate in 0.5 ml double distilled water. The resulting concentration is calculated using the lot-specific information, given on the label (final conc. approx 10 ng/ml). This solution is used for the preparation of hGH standards for establishing a hGH calibration curve (see 7.2: preparation of hGH standards).
Solution 2 Anti-hGH-DIG (bottle 2)
Reconstitute the lyophilizate in 0.5 ml double distilled water (final conc. 100 μg/ml).
Solution 2a Anti-hGH-DIG, working dilution
To prepare anti-hGH-DIG working dilution, dilute the reconstituted anti-hGH-DIG solution (100 g/ml) with sample buffer (solution 7) to a final conc. of 1 μg/ml (e.g., 100 μl of reconstituted anti-hGH-DIG solution 9.9 ml of solution 7 for 50 wells).The working dilution should be prepared freshly before use and should not be stored.
Solution 3 Anti-DIG-POD (bottle 3)
Reconstitute the lyophilizate in 0.5 ml double distilledwater (final conc. 20 U/ml). Do not add sodiumazide!
Solution 3a Anti-DIG-POD, working dilution
To prepare anti-DIG-POD, working dilution, dilute the reconstituted anti-DIG-POD solution (20 U/ml) with sample buffer (solution 7) to a final conc. of 200 mU/ml (e.g., 100 μl of reconstituted anti-DIG-POD solution 9.9 ml of solution 7 for 50 wells). The working dilution should be prepared freshly before use and should not be stored.
Solution 4 POD Substrate (bottle 4)
Ready-to-use solution.
Solution 5 POD Substrate containing Substrate Enhancer (bottles 4 and 5)
If a low hGH concentration in the sample is expected, add 1 mg of substrate enhancer (bottle 5) per ml of solution 4 and mix by stirring for 30 min at 15 to 25 °C. The solution is stable for only 2 hours and should therefore be prepared immediately before use. Use the substrate enhancer only if the hGH concentration is low!
Solution 6 Washing Buffer, 1× (bottle 6)
To prepare a ready-to-use washing buffer, mix 1 part 10x washing buffer concentrate (bottle 6) with 9 parts of double distilled water.
Solution 7 Sample Buffer (bottle 7)
The solution is ready-to-use. Mix thoroughly before use. Do not add sodium azide.
Storage conditions (working solution): Solution 1 hGH Stock Solution (bottle 1)
The reconstitutedsolution is stable for 2 months at 2 to 8 °C. For long term storage, store in aliquots at or below -15 to -25 °C.
Solution 2 Anti-hGH-DIG (bottle 2)
The reconstituted solution is stable for two months at 2 to 8 °C. For long term storage, store in aliquots at or below -15 to -25 °C.
Solution 2a Anti-hGH-DIG, working dilution
The working dilution should be prepared freshly before use and should not be stored.
Solution 3 Anti-DIG-POD (bottle 3)
The reconstituted solution is stable for 6 months at 2 to 8 °C. Do not freeze!
Solution 3a Anti-DIG-POD, working dilution
The working dilution should be prepared freshly before use and should not be stored.
Solution 4 POD Substrate (bottle 4)
The solution is stable until the expiration date given on the label if stored at 2 to 8 °C.
POD Substrate containing Substrate Enhancer (bottles 4 and 5)
The solution is stable for only 2 hours and should therefore be prepared immediately before use.
Solution 6 Washing Buffer, 1× (bottle 6)
The reconstituted solution is stable for 6 months at 2 to 8 °C.
Solution 7 Sample Buffer (bottle 7)
After opening the bottle we recommend storing the solution in aliquots at -15 to -25 °C, since it does not contain a preservative agent.

Analysis Note

No cross-reaction with rat growth hormone. Cross reactivity with the human homologues TSH, FSH, and LH has not been tested.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • hGH (human growth hormone)

  • Anti-hGH-digoxigenin (anti-hGH-DIG) antibody, lyophilizate

  • Anti-Digoxigenin-POD antibody, lyophilizate

  • POD Substrate ready-to-use

  • Substrate Enhancer

  • Washing Buffer 10x concentrated

  • Sample Buffer

  • Microplates (24x8 wells), precoated with anti-hGH (2)

  • Self-adhesive Plate Cover Foils (6)

See All (9)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jessica R Chaffey et al.
Scientific reports, 11(1), 15624-15624 (2021-08-04)
The generation of a human pancreatic beta cell line which reproduces the responses seen in primary beta cells, but is amenable to propagation in culture, has long been an important goal in diabetes research. This is particularly true for studies
Gabriela da Silva Xavier et al.
The Biochemical journal, 371(Pt 3), 761-774 (2003-02-19)
AMP-activated protein kinase (AMPK) has recently been implicated in the control of preproinsulin gene expression in pancreatic islet beta-cells [da Silva Xavier, Leclerc, Salt, Doiron, Hardie, Kahn and Rutter (2000) Proc. Natl. Acad. Sci. U.S.A. 97, 4023-4028]. Using pharmacological and
Woo-Jae Park et al.
Neuroscience letters, 444(3), 240-244 (2008-09-02)
Leucine-rich glioma inactivated 3 (LGI3) is a member of LGI/epitempin (EPTP) family. The biological function of LGI3 and its association with disease are not known. We previously reported that mouse LGI3 was highly expressed in brain in a developmentally and
Chelsea E Stamm et al.
mSphere, 4(3) (2019-06-07)
Mycobacterium tuberculosis (Mtb), the causative agent of tuberculosis, is one of the most successful human pathogens. One reason for its success is that Mtb can reside within host macrophages, a cell type that normally functions to phagocytose and destroy infectious

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service