Skip to Content
Merck
All Photos(2)

Key Documents

11277073910

Roche

DIG RNA Labeling Mix

sufficient for 20 reactions, solution

Synonym(s):

nucleic acid labeling

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105500
Pricing and availability is not currently available.

form

solution

Quality Level

usage

sufficient for 20 reactions

packaging

pkg of 40 μL

manufacturer/tradename

Roche

impurities

Ribonuclease, none detected (up to 20 µl using MSII-RNA)

color

colorless

solubility

water: miscible

storage temp.

−20°C

General description

Labeling efficiency: Approximately 10μg of full-length digoxigenin-labeled RNA is transcribed from 1μg linear template DNA.
Assay Time: 135 minutes
Sample Materials
Linearized plasmid DNA:
The DNA to be transcribed is cloned into the polylinker site of an appropriate transcription vector which contains adjacent to the polylinker a promoter for SP6, T7 or T3 RNA polymerase. For the synthesis of ‘run off′ transcripts the plasmid is linearized by a restriction enzyme. Restriction enzymes creating 5′-overhangs should be used; 3′ overhangs should be avoided. The linearized template DNA should be purified by phenol chloroform extraction and ethanol precipitation, to avoid RNase contamination. For ′run around′ transcription circular plasmid DNA is used.
PCR product:
PCR-fragments which contain RNA polymerase promoter sequences can also be used as templates for transcription. Purification of the PCR fragment by HighPure column purification prior to transcription is recommended.
DIG-labeled, single-stranded RNA probes of defined length are generated by in vitro transcription. DIG-11-UTP is incorporated by SP6, T7, and T3 RNA polymerases at approximately every 20 to 25th nucleotide of the transcript under standard conditions. The DIG RNA Labeling Mix is specifically designed for the use with SP6, T7, and T3 RNA polymerases, which are supplied with an optimized transcription buffer.
Convenient nucleotide mixture for the labeling of RNA with Digoxigenin-11-UTP.

Contents
10x solution with:
10 mM ATP, CTP, GTP (each), 6.5 mM UTP, 3.5 mM DIG-11-UTP.

Specificity

Heat inactivation: Stop the reaction by adding 2 μl 0.2 M EDTA (pH 8.0).

Application

RNA labeling with Digoxigenin-11-UTP by in vitro transcription with SP6, T7, and T3 RNA polymerases. DIG-labeled RNA is used in a variety of hybridization techniques:
  • Northern blots[1]
  • Southern blots
  • Dot blots
  • Plaque or colony lifts
  • RNase protection experiments
  • Chromosomes, cells, and tissue sections in situ[2][3]

Features and Benefits

The DIG RNA Labeling Mix is especially designed for the use with SP6,T7 and T3 RNA polymerases from Roche which are supplied with an optimized transcription buffer.

Quality

Function tested in the DIG RNA Labeling Kit and in the DIG Nucleic Acid Detection Kit.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ya Zhang et al.
Open biology, 10(9), 200172-200172 (2020-09-09)
Somatostatin (SS) and allatostatin-C (ASTC) are structurally and evolutionarily related neuropeptides that act as inhibitory regulators of physiological processes in mammals and insects, respectively. Here, we report the first molecular and functional characterization of SS/ASTC-type signalling in a deuterostome invertebrate-the
Ryan B MacDonald et al.
Developmental dynamics : an official publication of the American Association of Anatomists, 239(8), 2298-2306 (2010-07-27)
The Dlx genes encode a family of transcription factors important for the development of the vertebrate forebrain. These genes have very similar expression domains during the development of the telencephalon in mice and play a role in gamma-aminobutyric acid (GABAergic)
Nathalie Bessodes et al.
PLoS genetics, 8(12), e1003121-e1003121 (2012-12-29)
During echinoderm development, expression of nodal on the right side plays a crucial role in positioning of the rudiment on the left side, but the mechanisms that restrict nodal expression to the right side are not known. Here we show
Marco Nousch et al.
Journal of cell science, 126(Pt 18), 4274-4285 (2013-07-12)
Post-transcriptional regulatory mechanisms are widely used to control gene expression programs of tissue development and physiology. Controlled 3' poly(A) tail-length changes of mRNAs provide a mechanistic basis of such regulation, affecting mRNA stability and translational competence. Deadenylases are a conserved
Jiabin Chen et al.
Cerebral cortex (New York, N.Y. : 1991), 20(3), 650-660 (2009-07-03)
Experience-dependent plasticity of the adult visual cortex underlies perceptual learning and recovery of function following central nervous system lesions. To reveal the signal transduction cascades involved in adult cortical plasticity, we utilized a model of remapping of cortical topography following

Articles

Digoxigenin (DIG) labeling methods and kits for DNA and RNA DIG probes, random primed DNA labeling, nick translation labeling, 5’ and 3’ oligonucleotide end-labeling.

Digoxigenin (DIG) labeling methods and kits for DNA and RNA DIG probes, random primed DNA labeling, nick translation labeling, 5’ and 3’ oligonucleotide end-labeling.

Digoxigenin (DIG) labeling methods and kits for DNA and RNA DIG probes, random primed DNA labeling, nick translation labeling, 5’ and 3’ oligonucleotide end-labeling.

Digoxigenin (DIG) labeling methods and kits for DNA and RNA DIG probes, random primed DNA labeling, nick translation labeling, 5’ and 3’ oligonucleotide end-labeling.

Protocols

Determine the labeling efficiency in terms of μg (expected yield of a standard labeling reaction is 20 μg of DIG labeled RNA per μg linearized template DNA after the DIG RNA labeling reaction).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service