Skip to Content
Merck
All Photos(1)

Key Documents

11179179001

Roche

Collagen

from rat tail tendon

Synonym(s):

collagen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

biological source

rat tail

Quality Level

sterility

sterile

form

lyophilized (clear, colorless solution after reconstitution)

packaging

pkg of 30 mg

manufacturer/tradename

Roche

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

storage temp.

2-8°C

Gene Information

General description

Collagens are abundantly present in vertebrates. 41 specific genes codes for 42 polypeptide chains and produce 27 collagen types. Collagens  are mainly present in the extracellular matrix. Collagen type I is abundantly present in mammals. Type IV collagen is an important structural protein of basement membranes. It is chemically and genetically different from stroma collagen types I and III and cartilage collagen type II.

Specificity

Active on most mammalian cells.

Application

Collagen has been used in the preparation of coverslip coating solution and collagen gel contraction assay.
Collagen is used as a substrate for culturing a variety of cells, such as coating of culture dishes and the preparation of collagen gels.

Biochem/physiol Actions

Collagen provides extracellular support for multicellular animals. Collagen type I offers mechanical stability, strength and toughness to a range of tissues from tendons and ligaments, to skin, cornea, bone and dentin.

Features and Benefits

Contents
Lyophilizate, cell culture grade, 3 vials of 10mg

Specifications

Biological activity: Tested for the promotion of adherence of HUV-EC cells.

Preparation Note

Working concentration: 5 μg/cm2
5 μg/cm2 for the coating of cell culture vessels; for the preparation of collagen gels, a final concentration of 2 to 3 mg/ml is used.

Reconstitution

IFor best results, dissolve the lyophilizate in 0.2% acetic acid (v/v) solution.
For the preparation of collagen gels, the content of the bottle should be dissolved in 3.3 ml sterile 0.2% acetic acid (v/v) each. This results in a final concentration of 3 mg/ml.
For coating culture dishes the final concentration should be 1 to 2 mg/ml.
Note: For dissolving: do not stir, just pour the acetic acid onto the lyophilizate and allow it to stand for several hours until it has dissolved. To completely dissolve the product an incubation for up to a maximum of 24 hours at 15 to 25 °C may be required.

Reconstitution of the Collagen should be done at 15 to 25 °C, higher temperatures destroy the fibres and may prevent the subsequent gelation of the Collagen.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metastatic potential correlates with enzymatic degradation of basement membrane collagen.
Liotta L A, et al.
Nature, 284(5751), 67-67 (1980)
Collagen: Structure and Mechanics (2008)
Myelination of rodent hippocampal neurons in culture.
Gardner A, et al.
Nature Protocols, 7(10), 1774-1774 (2012)
Lodish H
Molecular and Cellular Biology (2000)
Identification and analysis of a novel dimerization domain shared by various members of JNK scaffold proteins.
Cohen-Katsenelson K, et al.
The Journal of Biological Chemistry (2013)

Articles

The extracellular matrix (ECM) is secreted by cells and surrounds them in tissues.

The extracellular matrix (ECM) is secreted by cells and surrounds them in tissues.

The extracellular matrix (ECM) is secreted by cells and surrounds them in tissues.

The extracellular matrix (ECM) is secreted by cells and surrounds them in tissues.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service