Skip to Content
Merck
All Photos(1)

Key Documents

11051482001

Roche

dTTP

Lithium salt

Synonym(s):

Thymidine 5′-triphosphate sodium salt, 2′-deoxy-thymidine-5′-triphosphate, deoxy-thymidine-5′-triphosphate, 2′-, TTP, dTTP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N2O14P3Na
CAS Number:
Molecular Weight:
504.15
MDL number:
UNSPSC Code:
41116100
PubChem Substance ID:

Quality Level

Assay

≥98%

form

solution

usage

sufficient for 6,250 standard reactions (PCR)

packaging

pkg of 250 μL (25 μmol)

manufacturer/tradename

Roche

concentration

100 mM

technique(s)

DNA amplification: suitable
DNA sequencing: suitable

storage temp.

−20°C

SMILES string

[NaH].[P](=O)(O[P](=O)(OCC1OC(CC1O)N2C=C(C(=O)NC2=O)C)O)(O[P](=O)(O)O)O

InChI

1S/C10H17N2O14P3.Na.H/c1-5-3-12(10(15)11-9(5)14)8-2-6(13)7(24-8)4-23-28(19,20)26-29(21,22)25-27(16,17)18;;/h3,6-8,13H,2,4H2,1H3,(H,19,20)(H,21,22)(H,11,14,15)(H2,16,17,18);;

InChI key

KAELIXTVSANDSE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

dTTP is a 100 mM clear, colorless solution of lithium salt (pH 7).
dTTP solution in water (pH 7), 100 mM, lithium salt.

Application

Use dTTP, lithium salt, for DNA sequencing, labeling, and all types of amplification reactions or primer-extension reactions.
It has been used in PCR fingrprinting and PCR techniques.

Quality

Typical analysis:
  • ≥98% dTTP (HPLC), ≤1.5% dTDP (HPLC), ≤0.5% dTMP (HPLC)
  • Absorbance determination A250/A260, A280/A260, A290/A260
  • DNase activity negative, according to the current Quality Control procedures
  • Suitability for PCR

Unit Definition

1 μmol dTTP = 9.5 A260units

Other Notes

For life science research only. Not for use in diagnostic procedures.

Our PCR Grade deoxynucleotides are even purer than the lithium salts, to ensure excellent performance in your PCRs. PCR Grade deoxynucleotides contain the sodium salt of the nucleotide in PCR Grade Water.
For your convenience, some of our deoxynucleotides are also available in:
  • nucleotide sets (all 4 nucleotides in separate vials)
  • ready-to-use nucleotide mixes (premixed solutions of all 4 nucleotides)

They are also vital components of:
  • kits (dNTPs, buffer, and polymerase in separate vials)
  • ready-to-use mixes [polymerase, nucleotides, and buffer (premixed 2x solutions)]

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Pillay et al.
Journal of clinical microbiology, 42(8), 3853-3856 (2004-08-07)
An immunochromatographic strip test, Xenostrip-Tv, was compared to wet mount and PCR for the diagnosis of Trichomonas vaginalis infection in women. Of 428 specimens tested, 54 (12.6%) were positive by an "expanded gold standard," defined as either a positive wet
Luciana Trilles et al.
Memorias do Instituto Oswaldo Cruz, 103(5), 455-462 (2008-09-18)
The molecular types of 443 Brazilian isolates of Cryptococcus neoformans and Cryptococcus gattii were analyzed to determine their geographic distribution within Brazil and their underlying host conditions. The following data, imported from previous epidemiological studies as well as two culture
Wieland Meyer et al.
Emerging infectious diseases, 9(2), 189-195 (2003-02-27)
A network was established to acquire basic knowledge of Cryptococcus neoformans in IberoAmerican countries. To this effect, 340 clinical, veterinary, and environmental isolates from Argentina, Brazil, Chile, Colombia, Mexico, Peru, Venezuela, Guatemala, and Spain were typed by using M13 polymerase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service