Skip to Content
Merck
All Photos(5)

Documents

ZLXE0150WW

Milli-Q

Elix® Essential Water Purification System

Type 2 pure water of consistent and reliable quality from potable tap water with a flow rate of 15 L/h

Synonym(s):

Elix® Essential 15 Water Purification System

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104202
NACRES:
JA.13

AC/DC input

100 - 230 V AC, 50 - 60 Hz (per your country requirements)

Quality Level

product line

Elix® Essential

packaging

pack of 1 unit

parameter

15 L/hr flow rate
17.2-18.5 kg operating weight (46.8-49.5 lb)
300 L/day max. usage (water)

resistivity

>5 MΩ-cm, at 25°C (Typically 10 - 15 MΩ.cm)

H × W × D

47 cm (18.5 in.) × 26.8 cm (10.6 in.) × 33.9 cm (13.3 in.)

input

feed water nature potable tap water

output

product water quality: type 2 water (> 5 MΩ·cm)

conductivity

<0.2 μS/cm at 25 °C (water output)

Looking for similar products? Visit Product Comparison Guide

General description

Elix® Essential delivers Type 2 pure water to support a broad range of research areas. It is designed to be fed from potable water and is the ideal feed for laboratory equipment, including ultrapure water systems.
In the Elix® Essential system′s water purification sequence, potable tap water is first treated with a Progard® pretreatment pack, followed by reverse osmosis (RO) to produce RO Type 3 water. This water enters the Elix® electrodeionization (EDI) module, which continuously regenerates ion-exchange resins using a small electrical field. This process requires only very small amounts of water and energy and results in consistently high-quality pure Type 2 water without a need for external chemical regeneration of the resin beads.
The pure water stored in a polyethylene (PE) reservoir can be either sourced through the tank front valve or sent by a distribution pump to feed laboratory instruments.

Application

Type 2 pure water is one of the most widely used reagents in the laboratory. It is used by scientists, researchers, and engineers everywhere in environments as diverse as academic, hospital, and quality control laboratories.

It is used as feed to laboratory equipment (e.g., Milli-Q® Type 1 ultrapure water systems, weathering testing chambers, autoclaves, glassware washers, and dissolution testing units). It is also used in the lab to prepare microbiological media, buffers and pH solutions, and solutions for use in histology. Examples of uses include thermodynamic studies, emulsions and membrane science.

Features and Benefits

  • Intelligent Reverse Osmosis (RO) controls water consumption, ensures a constant product flow rate (15 L/h).
  • Patented EDI module provides superior quality pure water while preserving the environment (self-regeneration of resin beads, minimal electricity consumption and no extra softener required).
  • Pure water stored in a PE tank minimizes risks of contamination during water storage.
  • The tank vent filter provides effective protection against airborne contaminants.
  • The ASM, Automatic Sanitization Module, prevents biofilm formation.
  • Resistivity values are >5 MΩ·cm at 25 °C, and TOC (Total Organic Carbon) <30 ppb.
  • The product water is of better quality than double-distilled water.
  • The Progard® cartridge incorporates RFID technology. This provides instant visibility on cartridge parameters and also ensures a safe and optimal performance of your system, giving you full traceability.
  • A color-coded backlit LCD clearly shows message importance with system alert and alarm icons.
  • Millitrack e-Solutions provides enhanced data management control, remote access capabilities to the system dashboard, and long-term electronic archiving for the water system.
  • With its small footprint, the system can be placed on or under the bench or wall-installed, depending on your needs, saving lab space.
  • Delivered with a Certificate of Calibration for the built-in temperature and resistivity meters, and a Certificate of Conformity ensuring that it has been built and tested fully assembled following internal Standard Operating Procedures (SOP).

Components

  • This is a web reference. The end letters "WW" will be replaced by the country letters on your invoice to have the right shipping kit for the country. This ensures you receive the user manual in the local language and a power cord adapted to the local electrical network.
  • The indicated price currently corresponds to the above catalogue number only; it is subject to change at any time without notice.
  • This catalogue number does not include consumables, accessories and other products and/or services that may be required for the installation and the correct operation of the water purification unit over time.
  • Because our customers each have specific purified water needs, we recommend that you have your purchase validated by one of our specialists. We remain at your disposal to advise you, do not hesitate to contact us!

Other Notes

For more information on this product please contact our Technical Service Team

Legal Information

ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLITRACK is a trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
PROGARD is a registered trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In-situ construction of antifouling separation layer via a reaction enhanced surface segregation method.
He, M. et al.
Chemical Engineering Science, 190, 89-97 (2018)
Improvement of the physical stability of oil-in-water nanoemulsions elaborated with Sacha inchi oil employing ultra-high-pressure homogenization.
Rave, M. C et al.
Journal of Food Engineering, 273, 109801-109801 (2020)
Manipulating membrane surface porosity and pore size by in-situ assembly of Pluronic F127 and tannin.
He, M. et al.
Journal of Membrane Science, 556, 285-292 (2018)
Solid-liquid equilibrium in the system 2-keto-L-gulonic acid+ sodium-2-keto-L-gulonate+ water.
Jirasek, F et al.
Fluid Phase Equilibria, 473, 318-322 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service