Skip to Content
Merck
All Photos(1)

Documents

VHPPWRSPCN

Millipore

Milliflex oasis® VHP resistant pump power supply

Milliflex Oasis® VHP pump power supply for CN

Synonym(s):

Milliflex Oasis® VHP pump power supply - CN, To connect up to 2 filtration pumps

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409

packaging

pkg of 1 ea

manufacturer/tradename

Milliflex®

technique(s)

Rapid Sterility Testing: suitable

application(s)

bioburden testing
pharmaceutical
sterility testing

compatibility

for use with Milliflex Oasis® VHP vacuum pump

Looking for similar products? Visit Product Comparison Guide

General description

Milliflex Oasis® filtration system offers a solution to simplify and secure microbial testing process with premium levels of productivity, result reliability and regulatory compliance. Milliflex Oasis® VHP resistant pump is a new generation vacuum pump for sterility and bioburden analysis in pharmaceutical environments. A power supply of European standard with 100 - 240 V, 50/60 enables efficient functioning of the filtration pump. One set of Milliflex Oasis® power supply can be used to power up to two pumps.

Application

Milliflex Oasis® VHP resistant pump finds its application in bioburden testing and sterility testing for Pharmaceutical Products Analysis.

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service