Key Documents
TANKVNT22
Milli-Q
Vent Filter
pore size 0.22 μm, carbon, and soda lime, Advanced protection against airborne contaminants for storage tanks
About This Item
UNSPSC Code:
41104212
NACRES:
NB.24
Quality Level
usage
(for type 3 water)
packaging
pkg of 2 units
manufacturer/tradename
Milli-Q®
H
33 cm (12.8 in.)
pore size
0.22 μm pore size
compatibility
for use with SDS
for use with tanks
General description
  • The vent filter protects highly sensitive stored pure water from possible contaminants in the environment. These include carbon dioxide, particles, micro-organisms and volatile organic compounds that mainly come from the laboratory atmosphere.
  • Contain a combination of purification media carefully selected to remove airborne contaminants that could degrade purified water quality during storage. Contains soda lime to prevent CO2 contamination.
  • The vent filter cartridges are constructed from polypropylene, polyethersulfone and styrene-acrylonitrile resin.
Application
Vent filter provides advanced protection against airborne contaminants.
Features and Benefits
Soda Lime for carbon dioxide removal.
Legal Information
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany