Skip to Content
Merck
All Photos(1)

Key Documents

SX0770

Supelco

Sodium Sulfide, Nonahydrate

meets analytical specifications of USP/NF GR ACS

Synonym(s):

Sodium Sulfide, Nonahydrate, Sodium Monosulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na2S · 9H2O
CAS Number:
Molecular Weight:
240.18
MDL number:
UNSPSC Code:
12352302
EC Index Number:
215-211-5
grade:
ACS reagent
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Agency

meets analytical specifications of USP/NF GR ACS

Assay

≥98.0% (iodometric)

color

colorless to slightly yellow

cation traces

Fe:, passes test
NH4+: ≤0.005%

shipped in

ambient

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[S-2].O.O.O.O.O.O.O.O.O

InChI

1S/2Na.9H2O.S/h;;9*1H2;/q2*+1;;;;;;;;;;-2

InChI key

ZGHLCBJZQLNUAZ-UHFFFAOYSA-N

General description

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Ammonium (NH4)0.005% maxAppearanceCrystals, colorless or only slight yellow colorAssay (iodometric)98.0% minIronTo pass testSulfite and thiosulfate (as SO2)0.1% max

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service