Skip to Content
Merck
All Photos(2)

Key Documents

SCGP00525

Millipore

Millipore® Steriflip® Vacuum Tube Top Filter

pore size 0.22 μm, polyethersulfone membrane, funnel capacity 50 mL, pack of 25 ea

Synonym(s):

Steriflip-GP Sterile Centrifuge Tube Top Filter Unit, tube top sterile filter unit, tube top vacuum filter, tube top vacuum sterile filtering unit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104914
eCl@ss:
32031602
NACRES:
NB.22

material

acrylic housing
polyethersulfone membrane
polypropylene

Quality Level

sterility

sterile

product line

Steriflip®

feature

holdup volume 0.6 mL
hydrophilic

packaging

pack of 25 ea

parameter

45 °C max. temp.

technique(s)

sterile filtration: suitable

H

138 mm

diam.

6.4 cm

filter diam.

40 mm

filtration area

7 cm2

funnel capacity

50 mL

working volume

50 mL

matrix

Express PLUS

pore size

0.22 μm pore size

fitting

double lead thread inlet (with vacuum port)
double lead thread outlet for 50 mL centrifuge tube

shipped in

ambient

General description

Compatible with 50 mL centrifuge tubes, Steriflip® filters eliminate the liquid transfer step by integrating directly into the sample tube. To filter, attach the unit to the 50 mL centrifuge tube containing your sample, flip, and apply vacuum. The filtrate collects directly into the attached 50 mL tube.

This Steriflip® 0.22 μm pore size polyethersulfone (PES) membrane filter unit combines a 50 mL centrifuge tube with a cap for processing and storage.

Application

For Research Use Only

Steriflip® filters are designed to be used for sterile vacuum filtration of aqueous solutions such as tissue culture media and biological fluids.
  • Tissue culture media +/- additives
  • Buffers
  • Biological solutions

Features and Benefits

  • Eliminates messy sample transfer via a simple vacuum-assisted, closed system
  • Can isolate cells from cellular debris using any of the various pore-sized nylon net filters
  • Ideal for sterilizing small batches of culture media
  • Attaches to any standard 50 mL centrifuge tube, eliminating unnecessary materials, time and risk of spillage

Legal Information

EXPRESS PLUS is a trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Steriflip is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christoph Ratzke et al.
Nature ecology & evolution, 4(3), 376-383 (2020-02-12)
Organisms-especially microbes-tend to live together in ecosystems. While some of these ecosystems are very biodiverse, others are not, and while some are very stable over time, others undergo strong temporal fluctuations. Despite a long history of research and a plethora
Jean-Christophe Cocuron et al.
Plant physiology, 181(3), 961-975 (2019-09-19)
Enhancing fatty acid synthesis (FAS) in maize (Zea mays) has tremendous potential nutritional and economic benefits due to the rapidly growing demand for vegetable oil. In maize kernels, the endosperm and the embryo are the main sites for synthesis and
Li Qiao et al.
Theranostics, 10(8), 3474-3487 (2020-03-25)
Cancer is the second leading cause of death worldwide and patients are in urgent need of therapies that can effectively target cancer with minimal off-target side effects. Exosomes are extracellular nano-shuttles that facilitate intercellular communication between cells and organs. It

Articles

Overview on how to maintain proper cell culture laboratory safety including steps on performing lab risk assessments, surface disinfection, biohazard and waste removal. Free ECACC handbook download.

Overview on how to maintain proper cell culture laboratory safety including steps on performing lab risk assessments, surface disinfection, biohazard and waste removal. Free ECACC handbook download.

Overview on how to maintain proper cell culture laboratory safety including steps on performing lab risk assessments, surface disinfection, biohazard and waste removal. Free ECACC handbook download.

Overview on how to maintain proper cell culture laboratory safety including steps on performing lab risk assessments, surface disinfection, biohazard and waste removal. Free ECACC handbook download.

Protocols

Purification protocols for immunoglobulins generally include affinity binding to Protein A or G chromatography media, washing unbound proteins, and eluting with a low pH buffer.

Best practices for standardizing and reducing the Extracellular Vesicle Preparation workflow, and eliminating contamination of EV preparations with dead-cell-derived vesicles.

Best practices for standardizing and reducing the Extracellular Vesicle Preparation workflow, and eliminating contamination of EV preparations with dead-cell-derived vesicles.

Best practices for standardizing and reducing the Extracellular Vesicle Preparation workflow, and eliminating contamination of EV preparations with dead-cell-derived vesicles.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service