Skip to Content
Merck
All Photos(2)

Documents

SCC066

Sigma-Aldrich

Blood-Brain Barrier hCMEC/D3 Cell Line

Human

Synonym(s):

Human BBB cell line, Human blood-brain barrier cell line, Human cerebral endothelial cell line, CMEC/D3 cell Line, hCMEC/D3 cell line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.81

product name

Blood-Brain Barrier hCMEC/D3 Cell Line, The hCMEC/D3 BBB cell line has been extensively characterized for brain endothelial phenotype and is a model of human blood-brain barrier (BBB) function.

biological source

human

Quality Level

technique(s)

cell culture | mammalian: suitable
drug transporter assay: suitable

shipped in

liquid nitrogen

General description

The blood–brain barrier (BBB) is a highly selective permeability barrier that separates the circulating blood from the brain extracellular fluid in the central nervous system. The blood–brain barrier is formed by capillary endothelial cells, which are connected by tight junctions with an extremely high electrical resistivity. The blood–brain barrier allows the passage of water, some gases, and lipid soluble molecules by passive diffusion, as well as the selective transport of molecules such as glucose and amino acids that are crucial to neural function. On the other hand, the blood–brain barrier may prevent the entry of lipophilic, potential neurotoxins by way of an active transport mechanism mediated by P-glycoprotein. Astrocytes are necessary to create the blood–brain barrier. The blood–brain barrier (BBB) prevents entry into the brain of most drugs from the blood. The presence of the BBB makes difficult the development of new treatments of brain diseases, or new radiopharmaceuticals for neuroimaging of brain.
The hCMEC/D3 cell line was derived from human temporal lobe microvessels isolated from tissue excised during surgery for control of epilepsy. The primary isolate was enriched in cerebral endothelial cells (CECs). In the first passage, cells were sequentially immortalized by lentiviral vector transduction with the catalytic subunit of human telomerase (hTERT) and SV40 large T antigen, following which CEC were selectively isolated by limited dilution cloning, and clones were extensively characterized for brain endothelial phenotype. This brain microvascular endothelial cell line represents one such model of the human BBB that can be easily grown and is amenable to cellular and molecular studies on pathological and drug transport mechanisms with relevance to the central nervous system (CNS).

Reference:
Couraud PO, et al. (2008) The human brain endothelial cell line hCMEC/D3 as a human blood‐brain barrier model for drug transport studies. Fluids Barriers CNS. 2013 Mar 26;10(1):16.

Cell Line Description

Endothelial Cells

Application

Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases
The hCMEC/D3 BBB cell line has been extensively characterized for brain endothelial phenotype and is a model of human blood-brain barrier (BBB) function.
This product is intended for sale and sold solely for internal non-commercial research use per the terms of the “Restricted Use Agreement” as detailed in the product documentation. For information regarding any other uses, please contact licensing@emdmillipore.com.

Quality

• Each vial contains ≥ 1X10^6 viable cells.
• Cells are tested by PCR and are negative for Hepatitis A, B, C, HPV, Herpes and HIV-1 & 2 viruses.
• Cells are negative for mycoplasma contamination.

Storage and Stability

Blood-Brain Barrier hCMEC/D3 cells should be stored in liquid nitrogen. The cells can be passage for at least 10 passages without significantly affecting the cell marker expression and functionality.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges {HCompany} to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kensuke Toyama et al.
Arteriosclerosis, thrombosis, and vascular biology, 38(6), 1392-1406 (2018-04-14)
There are currently no effective treatments for the prevention of dementia associated with vascular cognitive impairment. MicroRNAs regulate gene expression at the post-transcriptional level and play key roles in vascular disorders. TNFα (tumor necrosis factor-α) regulates blood-brain barrier breakdown through
Sophie Gong et al.
SLAS discovery : advancing life sciences R & D, 23(8), 832-841 (2018-03-06)
Antibody-triggered endocytosis (ATE) is a biological mechanism on which many therapeutic strategies are grounded, such as delivery of antibody-drug conjugates (ADCs). Current methods monitoring ATE include confocal Z-stack analysis, acid wash, antibody quenching, and pH-sensitive dye labeling. However, those generate
Rahul A Parikh et al.
Genes, chromosomes & cancer, 53(1), 25-37 (2013-10-22)
The ATR-CHEK1 pathway is upregulated and overactivated in Ataxia Telangiectasia (AT) cells, which lack functional ATM protein. Loss of ATM in AT confers radiosensitivity, although ATR-CHEK1 pathway overactivation compensates, leads to prolonged G(2) arrest after treatment with ionizing radiation (IR)
Lei Hao et al.
Croatian medical journal, 57(1), 51-57 (2016-03-05)
To explore the effects of hyperbaric oxygen preconditioning (HBOP) on the permeability of blood-brain barrier (BBB) and expression of tight junction proteins under hypoxic conditions in vitro. A BBB in vitro model was constructed using the hCMEC/D3 cell line and
Plasma Exosomes Disrupt the Blood-Brain Barrier in Children with Obstructive Sleep Apnea and Neurocognitive Deficits.
Abdelnaby Khalyfa et al.
American journal of respiratory and critical care medicine, 197(8), 1073-1076 (2017-10-21)

Related Content

Learn how to use our cell culture tested, ready-to-use MilliShot™ single dose antibiotic solutions, conveniently packaged in one-time use vials.

Learn how to use our cell culture tested, ready-to-use MilliShot™ single dose antibiotic solutions, conveniently packaged in one-time use vials.

Learn how to use our cell culture tested, ready-to-use MilliShot™ single dose antibiotic solutions, conveniently packaged in one-time use vials.

Learn how to use our cell culture tested, ready-to-use MilliShot™ single dose antibiotic solutions, conveniently packaged in one-time use vials.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service