PSMT010R
Millicell-24 Cell Culture Insert Plate
Synonym(s):
Cell Culture Insert Plate, Insert Plate for Cell Culture, Millicell-24 Plate
About This Item
Recommended Products
material
flat wells
polycarbonate membrane
polystyrene
Quality Level
description
Polycarbonate, 5 μm pore size, sterile, 24-well cell culture plate, single-well feeder tray, 24-well receiver tray and lid
sterility
sterile
feature
lid
manufacturer/tradename
Millicell®
parameter
50 °C max. temp.
technique(s)
cell attachment: suitable
cell culture | mammalian: suitable
cell differentiation: suitable
drug transporter assay: suitable
permeability assay: suitable
H
26.2 mm
L
128 mm
W
84.8 mm
filter diam.
9.5 mm
filtration area
0.7 cm2
plate size
24 wells
working volume
400 μL
matrix
Isopore™
pore size
5.0 μm pore size
binding type
Tissue Culture (TC)-treated surface
shipped in
ambient
General description
The Millicell device is available with a 0.4 µm PCF and a variety of PET membrane pore sizes. The plate is designed for maximum user convenience and incorporates features such as apical assist for easier pipetting and basolateral access. To reduce the risk of monolayer contamination, the membrane plates have feet to elevate the plate above the work surface when disassembled from the feeder tray. Tear drop-shaped receiver wells eliminate air bubbles as plates are assembled.
Millicell plate are optimized to grow and sustain high integrity cell monolayers. Cells grown on Millicell plate membranes grow better than on plastic because the cells are nourished from both the apical and basolateral sides. Cell growth and function more closely mimic in vivo. Millicell plates are designed for analysis as well as cell growth and can be used manually or with automated cell seeding, feeding and washing systems
With this complete system, you can grow, feed, and analyze cells in one membrane-bottom plate. The plate can be used with either a single-well or a multi-well feeding tray. At the time of transport analysis, the plate is simply transferred to a multi-well transport tray for analysis.
This streamlined design enhances functionality with:
- Seed-and-feed systems
- Automated liquid handling systems and span pipettes
- Basolateral and apical access to cells
- Transepithelial electrical resistance (TEER) measurement systems
Application
Packaging
Components
Physical form
Other Notes
5-pack excludes single-well trays product PSSW010R5.
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
This page covers the basic indirect co-culture procedure on both sides of Millicell cell culture insert membranes.
This page covers the basic indirect co-culture procedure on both sides of Millicell cell culture insert membranes.
This page covers the basic indirect co-culture procedure on both sides of Millicell cell culture insert membranes.
This page covers the basic indirect co-culture procedure on both sides of Millicell cell culture insert membranes.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service