Skip to Content
Merck
All Photos(1)

Key Documents

PC253L

Sigma-Aldrich

Anti-Calbindin D-28K (Ab-1) Rabbit pAb

lyophilized, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized

contains

≤0.1% sodium azide as preservative

species reactivity

baboon, monkey, rat, human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

ambient

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CALB1(793)
rhesus monkey ... Calb1(677721)

General description

Anti-Calbindin D-28K (Ab-1), rabbit polyclonal, reecognizes the Calbindin D-28K protein in rat hippocampus. It is validated for use in frozen sections and for T cell co-stimulation.
Rabbit polyclonal antibody supplied as lyophilized undiluted serum. Recognizes the calbindin D-28K protein.
Recognizes the Calbindin D-28K protein in rat hippocampus tissue.

Immunogen

Bovine
purified bovine cerebellum calbindin D-28K protein

Application

Frozen Sections (whole mount/vibrotome sections, 1:1000, fluorescence)

Vibrotome Sections (1:5000-1:10,000 for biotin-streptavidin/peroxidase detection, see application references)

Warning

Toxicity: Standard Handling (A)

Physical form

Undiluted serum.

Reconstitution

Reconstitute the lyophilized antibody with 100 µl sterile distilled H₂O. Resulting reconstituted solution will contain ≤0.1% sodium azide. Be careful to reconstitute the entire contents of the vial; during shipment and handling portions of the lyophilized pellet may have become dislodged and may not be in the bottom of the vial. Dilute with sterile PBS or Tris buffer at dilutions no higher than 1:10. Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C. Avoid freeze/thaw cycles of solutions.

Analysis Note

Positive Control
Rat striatum, hippocampus, or cortex

Other Notes

Antibody specificity was examined in the rat striatum, cortex, and hippocampus. Detection of primary antibody with biotin-streptavidin/peroxidase reagents according to manufacturers directions. Antibody should be titrated for optimal results in individual systems.
Conde, F., et al. 1994. J. Comp. Neurol.341, 95.
Heizmann, C.W. 1993. Acta Neurobiol. Exp.53, 15.
Baimbridge, K.G., et al. 1992. Trends Neurosci.15, 303.
Heizmann, C.W. and Hunziker, W. 1992. Trends Biochem. Sci.16, 98.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana I Arroyo et al.
EMBO molecular medicine, 6(3), 398-413 (2014-01-23)
Understanding the role of lipids in synapses and the aberrant molecular mechanisms causing the cognitive deficits that characterize most lipidosis is necessary to develop therapies for these diseases. Here we describe sphingomyelin (SM) as a key modulator of the dendritic
Daniela Herrera Moro Chao et al.
PloS one, 10(9), e0138107-e0138107 (2015-09-30)
Gaucher disease is characterized by lysosomal accumulation of glucosylceramide due to deficient activity of lysosomal glucocerebrosidase (GBA). In cells, glucosylceramide is also degraded outside lysosomes by the enzyme glucosylceramidase 2 (GBA2) of which inherited deficiency is associated with ataxias. The
B A Patel et al.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society, 22(8), 909-918 (2010-05-21)
Inhibitory neurotransmission to the longitudinal muscle is more prominent in the neonatal than in the adult guinea pig ileum. Inhibitory neuromuscular transmission was investigated using in vitro ileal longitudinal muscle myenteric plexus (LMMP) preparations made from neonatal (< or =48
André R A Marques et al.
PloS one, 10(8), e0135889-e0135889 (2015-08-15)
The enzyme glucocerebrosidase (GBA) hydrolyses glucosylceramide (GlcCer) in lysosomes. Markedly reduced GBA activity is associated with severe manifestations of Gaucher disease including neurological involvement. Mutations in the GBA gene have recently also been identified as major genetic risk factor for
Satoshi Watanabe et al.
PloS one, 8(1), e54146-e54146 (2013-01-22)
Adeno-associated virus (AAV) is well established as a vehicle for in vivo gene transfer into the mammalian retina. This virus is promising not only for gene therapy of retinal diseases, but also for in vivo functional analysis of retinal genes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service