Skip to Content
Merck
All Photos(1)

Documents

PC177L

Sigma-Aldrich

Anti-Serotonin (5-HT) Transporter (602-622) Rabbit pAb

lyophilized, Calbiochem®

Synonym(s):

Rabbit Anti-Serotonin Transporter, Serotonin Transporter Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized

contains

≤0.1% sodium azide as preservative

species reactivity

mouse, rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

ambient

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Slc6A4(15567)

Related Categories

General description

Rabbit polyclonal antibody supplied as lyophilized, undiluted serum. Recognizes the serotonin (5-HT) transporter protein.
Recognizes the serotonin (5-HT) transporter in hypothalamus raphe nuclei and spinal cord.
This Anti-Serotonin (5-HT) Transporter (602-622) Rabbit pAb is validated for use in Frozen Sections, Immunofluorescence for the detection of Serotonin (5-HT) Transporter (602-622).

Immunogen

Rat
a synthetic peptide corresponding to amino acids 602-622 of rat 5-HT transporter

Application


Frozen Sections (1:10,000-1:15,000; see application references)
Immunoblotting (see comments)
Immunofluorescence (1:200-1:400)

Warning

Toxicity: Standard Handling (A)

Physical form

Undiluted serum.

Reconstitution

Reconstitute the lyophilized antibody with 100 µl of sterile distilled H₂O. Reconstitute the entire contents of the vial; during shipment and handling portions of the lyophilized pellet may have become dislodged and may not be in the bottom of the vial. Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C. Further dilute with sterile PBS or Tris buffer to dilutions no higher than 1:10.

Analysis Note

Positive Control
Rat naphe nuclei, hypothalamus, cortex, or spinal cord

Other Notes

Antibody specificity was examined in the rat cortex, hypothalamus, raphe nuclei, and spinal cord. Staining is completely eliminated by pretreatment of antibody with Serotonin (5-HT) Transporter Control Peptide (Cat. No. PP87) at a concentration of 5 µg/ml. Low levels of glutaraldehyde (0.1%-0.3%) may be used in conjunction with paraformaldehyde for tissue perfusion fixation. The specificity of this antibody was also determined by immunoblotting analysis using rat brain extracts of the cortex, hypothalamus, midbrain, and hindbrain, which specifically detected a single band. Antibody should be titrated for optimal results in individual systems.
Zhou, F.C., et al. 1996. Mol. Brain Res.43, 267.
Blakely, R.D., et al. 1994. J. Exp. Biol.196, 263.
Coccaro, E.F. and Murphy, D.L. (Eds.) 1990. in Serotonin in Major Disorders. The Progress in Psychiatry Series, American Psychiatric Press, Washington, D.C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diana K Sarko et al.
The Journal of comparative neurology, 519(1), 64-74 (2010-12-02)
Northern grasshopper mice (Onychomys leucogaster) are among the most highly carnivorous rodents in North America. Because predatory mammals may have specialization of senses used to detect prey, we investigated the organization of sensory areas within grasshopper mouse neocortex and quantified
Rebecca L Lowery et al.
International journal of molecular sciences, 22(4) (2021-03-07)
While microglia have been established as critical mediators of synaptic plasticity, the molecular signals underlying this process are still being uncovered. Increasing evidence suggests that microglia utilize these signals in a temporally and regionally heterogeneous manner. Subsequently, it is necessary
R Ye et al.
Genes, brain, and behavior, 13(3), 261-275 (2014-01-11)
Presynaptic serotonin (5-hydroxytryptamine, 5-HT) transporters (SERT) regulate 5-HT signaling via antidepressant-sensitive clearance of released neurotransmitter. Polymorphisms in the human SERT gene (SLC6A4) have been linked to risk for multiple neuropsychiatric disorders, including depression, obsessive-compulsive disorder and autism. Using BXD recombinant
Andrea Forero et al.
Frontiers in cellular neuroscience, 11, 307-307 (2017-10-12)
Background: During early prenatal stages of brain development, serotonin (5-HT)-specific neurons migrate through somal translocation to form the raphe nuclei and subsequently begin to project to their target regions. The rostral cluster of cells, comprising the median and dorsal raphe
Ibrahim M Shokry et al.
BMC pharmacology & toxicology, 20(1), 47-47 (2019-08-07)
MDMA causes serotonin (5-HT) syndrome immediately after administration and serotonergic injury in a few days or weeks. However, a serotonin syndrome is not always followed by serotonergic injury, indicating different mechanisms responsible for two adverse effects. The goal of present

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service