Skip to Content
Merck
All Photos(1)

Key Documents

NS225

Sigma-Aldrich

Neurite Outgrowth Assay Kit (1 µm)

The NS225 Neurite Outgrowth Assay Kit (1 μm) is based on the use of Millicell cell culture inserts (chambers) containing a permeable membrane with 1 μm pores at the base.

Synonym(s):

Neurite growth assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®

technique(s)

cell based assay: suitable

detection method

colorimetric

shipped in

wet ice

General description

Application CHEMICON′s NS225 Neurite Outgrowth Assay Kit (1 μm) utilizes microporous tissue culture insert technology from Millipore and is based on the use of Millicell cell culture inserts (chambers) containing a permeable membrane with 1 μm pores at the base. The inserts are designed to fit into a receiver vessel, which when in use contains differentiation media in contact with the bottom of the insert membrane. The permeable membranes allow for discrimination between neurites and cell bodies, as projecting neurites will pass easily through the pores but cell bodies will not. Therefore, by inducing neurites to traverse these membrane pores, a purified population of neurites is located on the underside of the membrane, distal to the surface on which neural cell bodies are deposited. This kit has has been specifically designed for and validated with PC12 cells. The diameter of PC12 cell bodies is relatively small compared to that of many other cell types commonly used in neurite outgrowth assays [5, 6, 7]. As a result, many PC12 cell bodies may pass though pores larger than 1 μm, significantly obscuring neurite outgrowth assay results. Using a 1 μm pore size greatly reduces background and improves the signal-to-noise ratio in studies using PC12 cells. Use of this 1 μm pore size with larger cell types that extend wider diameter neurites, e.g., N1E-115 cells, will greatly diminish the number of neurites able to project through to the underside of the membrane, and is thus not recommended. The assay is a simple, efficient, and versatile system for the quantitative determination of compounds that influence neurite formation and repulsion. With this system, it is possible to screen numerous biological and pharmacological agents simultaneously, directly evaluate adhesion and guidance receptor functions responsible for neurite extension and repulsion, as well as analyze gene function in transfected cells. The microporous filter allows for biochemical separation and purification of neurites and cell bodies for detailed molecular analysis of protein expression, signal transduction processes and identification of drug targets that regulate neurite outgrowth or retraction processes. Reagents and materials supplied in the NS225 Neurite Outgrowth Assay Kit are sufficient for 12 tests.

Application

The NS225 Neurite Outgrowth Assay Kit (1 μm) is based on the use of Millicell cell culture inserts (chambers) containing a permeable membrane with 1 μm pores at the base.

Components

Neurite Outgrowth Plate Assembly, 1 µm: (Part No. 2007254) 1 x 24-well plate containing 12 x Millicell hanging inserts with 1 µm pore size membranes.

Neurite Stain Solution: (Part No. 90242) One 20 mL bottle.

Neurite Stain Extraction Buffer: (Part No. 90243) One 20 mL bottle.

Neurite Outgrowth Assay Plate: (Part No. 2007255) Two 24-well plates.

Cotton Swabs: (Part No. 10202) 50 swabs.

Forceps: (Part No. 10203) One each.

Storage and Stability

When stored at 2-8° C, the kit components are stable up to the expiration date. Do not freeze or expose to elevated temperatures. Discard any remaining reagents after the expiration date.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Lamoureux et al.
The Journal of cell biology, 110(1), 71-79 (1990-01-01)
Several groups have shown that PC12 will extend microtubule-containing neurites on extracellular matrix (ECM) with no lag period in the absence of nerve growth factor. This is in contrast to nerve growth factor (NGF)-induced neurite outgrowth that occurs with a
D G Drubin et al.
The Journal of cell biology, 101(5 Pt 1), 1799-1807 (1985-11-01)
Nerve growth factor (NGF) regulates the microtubule-dependent extension and maintenance of axons by some peripheral neurons. We show here that one effect of NGF is to promote microtubule assembly during neurite outgrowth in PC12 cells. Though NGF causes an increase
M T Filbin
Current biology : CB, 10(3), R100-R103 (2000-02-19)
Myelin is a potent inhibitor of axon regeneration, but has been viewed as just one of many factors that prevent regeneration after injury. So it comes as a surprise that immunization against myelin has been found to allow extensive axon
J Qiu et al.
Glia, 29(2), 166-174 (2000-01-14)
The lack of axonal regeneration in the adult mammalian CNS is due to both unfavorable environmental glial factors and the intrinsic neuronal state. Inhibitors associated with myelin and the glial scar have been extensively studies and it has been shown
Tomoyuki Abe et al.
Journal of cell science, 116(Pt 1), 155-168 (2002-11-29)
Rho family small GTPases regulate multiple cellular functions through reorganization of the actin cytoskeleton. Among them, Cdc42 and Tc10 induce filopodia or peripheral processes in cultured cells. We have identified a member of the family, designated as RhoT, which is

Articles

Optimized cell based neurite outgrowth assays and reagents to study neuron function and development.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service