NB20
Anti-AnkyrinG Mouse mAb (463)
liquid, ≥95% (SDS-PAGE), clone 463
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
463, monoclonal
Assay
≥95% (SDS-PAGE)
form
liquid
contains
≤0.1% sodium azide as preservative
species reactivity
rat, mouse, human, canine
manufacturer/tradename
Calbiochem®
storage condition
do not freeze
isotype
IgG1
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... ANK3(288)
General description
Immunogen
Application

Frozen Sections (1-2 g/ml)
Immunoblotting (2-5 g/ml)
Immunofluorescence (1-2 g/ml)
Packaging
Warning
Physical form
Analysis Note
Mouse or rat brain extract or unmyelinated axons
Other Notes
Kunimoto, M., et al. 1995. FEBS Lett.357, 217.
Lambert, S., et al. 1995. Mol. Biol. Cell6, 98a supplement.
Bennett, V. and Gilligan, D.M. 1993. Ann. Rev. Cell Biol.9, 27.
Kunimoto, M., et al. 1991. J. Cell Biol.115, 1319.
Otto, E., et al. 1991. J. Cell Biol.114, 241.
Davis, J., and Bennett, V. 1984. J. Biol. Chem.259, 13550.
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service