MZAAWG251
Microfil® with EZ-Pak®
sample volume 250 mL, mixed cellulose esters (MCE), pore size 0.8 μm, filter diam. 47 mm, white filter, gridded filter
Synonym(s):
Membrane filtration system
About This Item
Recommended Products
material
gridded filter
mixed cellulose esters (MCE)
polypropylene
white filter
Agency
ISO 7704
description
Yeast or mold
sterility
sterile; ethylene oxide treated
feature
easily wetted with water
hydrophilic
manufacturer/tradename
Microfil® with EZ-Pak®
parameter
250 mL sample volume
filter diam.
47 mm
funnel capacity
250 mL
matrix
MF-Millipore™
pore size
0.8 μm pore size
input
sample type beverage(s)
sample(s) (environmental)
sample type sport drink
sample type cosmetics
sample type bottled water
sample type wine
sample type soft drink
sample type cider
sample type beer
sample type food(s)
application(s)
bioburden testing
clinical
cosmetics
environmental
environmental
food and beverages
industrial qc
compatibility
for use with EZ-Fit® Filtration Manifold
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Principle
Physical form
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Microfil® filtration system ensures microbiological quality assurance in routine testing of raw materials.
Microfil® filtration system ensures microbiological quality assurance in routine testing of raw materials.
Microfil® filtration system ensures microbiological quality assurance in routine testing of raw materials.
Microfil® filtration system ensures microbiological quality assurance in routine testing of raw materials.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service