Skip to Content
Merck

MX0HC10FS1

Millipore

Millistak+® HC Pod Depth Filter

X0HC, filtration area 1.1 m², Process Scale

Synonym(s):

Millistak+ HC Pod Depth Filter, X0HC media series, 1.1 m 2 surface area

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151806
eCl@ss:
32031690
Pricing and availability is not currently available.

Product Name

Millistak+® HC Pod Depth Filter, X0HC, cellulose, filtration area 1.1 m2, hydrophilic

material

cellulose
filter aid (inorganic)
glass fiber-reinforced polypropylene

Quality Level

description

Secondary clarification of bioreactor harvests, primarily for cell cultures

feature

hydrophilic

manufacturer/tradename

Millistak+®

parameter

1 bar max. differential pressure (15 psid) at 80 °C (Forward)
1 bar max. inlet pressure (15 psi) at 80 °C
2.1 bar max. differential pressure (30 psid) at 25 °C (Forward)
2.1 bar max. differential pressure (30 psid) at 25 °C (Reverse)
3.5 bar max. inlet pressure (50 psi) at 25 °C

H

32 cm (12.5 in.)

L

62 cm (24.2 in.)

filtration area

1.1 m2

impurities

<0.25 EU/mL bacterial endotoxins (LAL test, Aqueous extraction)

input

sample type liquid

conductivity

19.34-53.2 μS/cm at 25 °C (product water)

filter grade

X0HC

Looking for similar products? Visit Product Comparison Guide

General description

The Millistak+® depth filter media is offered in a scalable, disposable format providing greater flexibility because of its unique modular design. The Millistak+® MX0HC10FS1 comes in a POD format featuring a 32 cm (H) x 62 cm (L) and a filtration area of 1.1 m2.

This X0HC media series delivers optimal performance through a gradient density matri, ideal for secondary clarification post depth filtration or centrifugation. Can also be utilized in downstream processing steps to protect chromatography columns. applications. Millistak+® Pod filters are available in three distinct series of media grades in order to meet your specific needs - DE, CE and HC.

The Millistak+® Pod system is ideal for a wide variety of primary and secondary clarification applications, including cell cultures, yeast and E. coli lysates post centrifuge, E. coli refolds, media, vaccines, plasma proteins and sera.

Easy to Use

With the compact, modular design of our Millistak+® new Pod system, you can increase productivity and shorten cycle times.

Installation and setup of the Pod system is simple and straightforward. The unique design of the disposable adapters and disposable manifolds makes it easy to connect the Pod filters to rest of the unit operations in the process. The self-contained and disposable nature of the system protects operators from exposure to biohazards and eliminates maintenance as well as cleaning validation requirements.

Application

Secondary clarification of bioreactor harvests, primarily for cell cultures

Features and Benefits

  • Low hold-up volume for greater product yield
  • Broad range of media types offered in single and multilayer products
  • Millistak+® HC dual-action media improves prefiltration and compresses clarification
  • Flexible, modular format offers scalability from 5 to 12,000 liters or more
  • Patented disposable design eliminates need for housing, CIP or cleaning validation
  • Self-contained Pod filters protect operators from exposure to biohazards
  • Robust construction is easy to use and set up
  • Smaller footprint facilitates use in tight spaces

Specifications

Filter Grade: X0HC
Per 24 hour soak in 10 L pure water post autoclave (1 cycle of 60 minutes at 123 °C) and pure water flush of 100 L/m2 (10 L/ft2 ) of media surface area:
Sodium: <14.1 mg/ft2
Calcium: <5.3 mg/ft2
Magnesium: <3.9 mg/ft2
Mercury: <0.001mg/ft2
Chromium: ≤0.001 mg/ft2
Aluminum: <0.084 mg/ft2
Potassium: <4.2mg/ft2
Lead: <0.001 mg/ft2
Arsenic: <0.009mg/ft2
Cobalt: ≤0.001 mg/ft2
Iron: <0.064 mg/ft2
Manganese: ≤ 0.124 mg/ft2
Titanium: <0.002 mg/ft2
Zinc: <0.046 mg/ft2
Nickel: <0.013
Copper: <0.009

Preparation Note

Sterilization Method
1 autoclave cycle of 60 min @ 123 ºC

Analysis Note

Product Water Conductivity at 25°C: post autoclave (1 cycle of 60 minutes at 123 °C) and pure water flush of 10 liters per ft2 (100 L/m2) of media surface area

Legal Information

MILLISTAK+ is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service