Skip to Content

MSNU030

MultiScreen-PCR96 Filter Plate

Purification of PCR products

Synonym(s):

Montage PCR Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41104923
NACRES:
NB.15
eCl@ss:
32039006
Material:
flat bottom wells, polystyrene
Pore size:
pore sizeproprietary
Sterility:
non-sterile
Binding type:
low binding surface
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

flat bottom wells, polystyrene

sterility

non-sterile

product line

MultiScreen®

feature

lid

parameter

100 μL sample volume (per well)

technique(s)

DNA purification: suitable, PCR: suitable, activity assay: suitable (with precipitations), ligand binding assay: suitable, receptor binding assay: suitable

plate size

96 wells

well maximum volume

300 μL

working volume

150-300 μL

pore size

pore sizeproprietary

application(s)

sample preparation

binding type

low binding surface

shipped in

ambient

General description

The polymerase chain reaction (PCR) is a highly effective fundamental technique in molecular biology. It offers a fast and efficient way to amplify specific DNA or RNA sequences from nucleic acids derived from different sources using enzymatic processes in a laboratory setting. We have an extensive range of PCR consumables and equipment designed to facilitate every stage of your PCR workflow MultiScreen-PCR96 Filter plates can be used to purify PCR products to be used in further downstream applications.

Application

MultiScreen-PCR96Filter Plate is suitable for use:

  • in receptor/ligand binding assays
  • in sample preparation
  • in enzyme assays with precipitation
  • in DNA purification
  • in polymerase chain reaction (PCR)
  • in purifying polymerase chain reaction (PCR) products for mutation analysis
  • in purifying PCR products for measuring CAG repeats
  • in purifying PCR products for sequencing

Features and Benefits

Proprietary membrane

Physical form

Color Code: Clear

Other Notes

Automation Compatible: Yes
Research Category
Assays
Nucleic acid
Protein
Does not include reagents

Legal Information

MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

10-13 - German Storage Class 10 to 13



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library