Skip to Content
Merck
All Photos(3)

Key Documents

MD0SP20CL3

Millipore

Millistak+® HC Pro Depth Filter in Micro 20 format

greener alternative

D0SP, filtration area 20 cm², Process Development Scale

Synonym(s):

20 cm2 surface area, Female-Luer fitting connection, Millistak+ D0SP Micro 20 Depth Filter, Millistak+ HC Pro Pod Depth Filter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151806
Pricing and availability is not currently available.

Product Name

Millistak+® HC Pro Depth Filter in Micro 20 format, D0SP, silica filter aid (with Polyacrylic fiber/ non-woven material), filtration area 20 cm2, hydrophilic

material

glass filled polypropylene housing
silica filter aid (with Polyacrylic fiber/ non-woven material)

Quality Level

feature

hydrophilic

manufacturer/tradename

Millistak+®

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

diam.

2.5 in. (6.4 cm)

filtration area

20 cm2

impurities

<0.25 EU/mL (LAL test, clot test technique (on filter media only))

input

sample type liquid

fitting

Luer fitting

filter grade

D0SP

greener alternative category

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Device Configuration: Micro 20
We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives . This product is a Design for Sustainability (DfS) developed product. Click here to view its DfS scorecard.

Application

Primary clarification (direct harvest)

Features and Benefits

Millistak+® HC Pro Micro 20 devices offer:

  • Improved scalability during media screening trials
  • Smaller filtration area slightly reduces feedstream volume requirements (approximately 15% less).
  • Transparent housing enables visibility of fluid flow
  • Easier to calculate flow rates based on 20 cm2 filter area


Millistak+® HC Pro media offers:
  • Synthetic materials of construction
  • Reduced TOC extractables and a 50% reduction in the recommended pre-use flush volumes
  • No beta glucans to interfere with limulous amoebocyte lysate (LAL) testing for bacterial endotoxins
  • Lot-to-lot consistency for successful development and implementation of robust clarification processes
  • Two times the filtration capacity with equivalent filter retention properties over commercial DE-based benchmarks
  • Improved HCP impurity clearance


Millistak+® Pod filters offer:
  • Low hold-up volume for greater product yield
  • Flexible, modular format offers scalability up to 20,000 liters
  • Patented disposable design eliminates need for housing, CIP or cleaning validation
  • Self-contained Pod filters protect operators from exposure to biohazards
  • Robust construction is easy to use and set up
  • Smaller footprint facilitates use in tight spaces

Packaging

3 EA

Other Notes

Integrity is maintained after 2 autoclave cycles of 60 minutes at 123 °C

Legal Information

MILLISTAK+ is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

MILLISTAK+® is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Learn more on mAb downstream processing, more specifically clarification and the relevant associated products.

Learn more on mAb downstream processing, more specifically clarification and the relevant associated products.

Learn more on mAb downstream processing, more specifically clarification and the relevant associated products.

Learn more on mAb downstream processing, more specifically clarification and the relevant associated products.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service