Skip to Content
Merck
All Photos(3)

Key Documents

MABT868

Sigma-Aldrich

Anti-acetyl-alpha tubulin Antibody, clone 6-11B-1

clone 6-11B-1, from mouse

Synonym(s):

Tubulin alpha-1 chain, acetyl-alpha tubulin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6-11B-1, monoclonal

species reactivity

sea urchin, bovine, monkey, Chlamydomonas, Drosophila, mouse, rat-kangaroo, human

technique(s)

dot blot: suitable
electron microscopy: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG2bκ

GenBank accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

acetylation (Lys40)

Gene Information

human ... TUBA1A(7846)

General description

Microtubules are cytoskeletal filaments composed of of alpha-/beta-tubulin heterodimers that self-assemble head-to-tail to form protofilaments and laterally to form a hollow tube. Various post-translational modifications (PTMs), including acetylation, polyglutamylation, polyglycylation, detyrosination, phosphorylation, and palmitoylation, can regulate the polymerization properties of tubulins and/or their interactions with microtubule associated proteins (MAPs) and motor proteins. A subset of cytoplasmic microtubules and microtubules in the spindle, axon, and cilia are known to contain alpha-tubulin acetylated via the epsilon-amino group of Lysine-40 (K40). The GNAT lysine acetyltransferase family member MEC-17 (or alpha-tubulin acetyltransferase/alphaTAT) catalyzes K40 acetylation, while HDAC6 and sirtuin2 (SIRT2) are known to catalyze tubulin deacetylation. Experimental evidences indicate that K40 acetylation primes alpha-tubulin for further PTM(s), which cannot be reversed by later deactylation of K40ac. Subsequently, the acetylated and deacetylated alpha-tubulin in native microtubules are structurally distinct from that of unacetylated alpha-tubulin (never before has its K40 acetylated).

Specificity

Experimental evidences indicate that the acetylated and deacetylated α-tubulin in native microtubules are structurally distinct from that of unacetylated α-tubulin, and that clone 6-11B-1 recognizes acetylated and deacetylated α-tubulin, but not unacetylated (never acetylated) α-tubulin. While 6-11B-1 immunostaining pattern resembles that of acetyl-Lys40 antibodies on normal cylcling cells, caution must be taken when interpreting immunostaining results using this mAb on cells subjected to tubulin deacetylation manipulations by exogenous deacetylase expression or via chemical means.

Immunogen

15 S dynein fraction from sea urchin sperm axoneme.

Application

Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)
This Anti-acetyl-alpha tubulin Antibody, clone 6-11B-1 is validated for use in Western Blotting, Immunocytochemistry, Dot Blot and Electron Microscopy for the detection of acetyl-alpha tubulin.
Western Blotting Analysis: 4.0 µg/mL from a representative lot detected acetyl-alpha tubulin in 10 µg of mouse brain tissue lysate.
Immunocytochemistry Analysis: A representative lot detected acetylated alpha-tubulin in cultured normal human kidney epithelial cells (Courtesy of Dr Christopher Ward, University of Kansas Medical Center).
Western Blotting Analysis: A representative lot detected alpha-tubulin in sea urchin sperm axoneme lysates, but not in lysates from taxol-stabilized microtubules from sea urchin eggs (Piperno, G., and Fuller, M. (1985). J Cell Biol.101(6):2085-2094).
Dot Blot Analysis: A representative lot detected alpha-tubulin in both the ionic detergent Sarkosyl-soluble and insoluble fractions from sea urchin spern axoneme extracts (Piperno, G., and Fuller, M. (1985). J Cell Biol.101(6):2085-2094).
Immunocytochemistry Analysis: A representative lot detected acetylated and deacetylated, but not unacetylated alpha-tubulin in COS-7 green monkey kidney fibroblasts and Ptk2 rat kangaroo kidney epithelial cells by fluorescent immunocytochemistry (Soppina V., et al. (2012). PLoS One. 7(10):e48204).
Electron Microscopy Analysis: The Fab fragment of clone 6-11B-1 has been shown to stain Taxol-stabilized microtubules polymerized in vitro from purified bovine brain tubulin with or without SIRT2-catalyzed deacetylation (Soppina V., et al. (2012). PLoS One. 7(10):e48204).

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 4.0 µg/mL of this antibody detected acetyl-alpha tubulin in 10 µg of HeLa cell lysate.

Target description

~50 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Markus Masek et al.
Nature communications, 13(1), 1282-1282 (2022-03-13)
Primary cilia are key sensory organelles whose dysfunction leads to ciliopathy disorders such as Bardet-Biedl syndrome (BBS). Retinal degeneration is common in ciliopathies, since the outer segments (OSs) of photoreceptors are highly specialized primary cilia. BBS1, encoded by the most
Tamas Yelland et al.
Journal of medicinal chemistry, 65(3), 1898-1914 (2022-02-03)
RAS is a major anticancer drug target which requires membrane localization to activate downstream signal transduction. The direct inhibition of RAS has proven to be challenging. Here, we present a novel strategy for targeting RAS by stabilizing its interaction with
Vincent Serapiglia et al.
Frontiers in pediatrics, 9, 780166-780166 (2022-03-15)
Fetal endoscopic tracheal occlusion (FETO) is an emerging surgical therapy for congenital diaphragmatic hernia (CDH). Ovine and rabbit data suggested altered lung epithelial cell populations after tracheal occlusion (TO) with transcriptomic signatures implicating basal cells. To test this hypothesis, we
Yan Shin J Liao et al.
Experimental physiology, 105(10), 1673-1683 (2020-08-01)
What is the central question of this study? What is the impact of airway cholinergic history on the properties of airway mucus secretion in a cystic fibrosis-like environment? What is the main finding and its importance? Prior cholinergic challenge slightly
Shin-Ping Kuan et al.
iScience, 19, 737-748 (2019-09-07)
Prolonged heat and sea salt aerosols pose a challenge for the mammalian airway, placing the protective airway surface liquid (ASL) at risk for desiccation. Thus, mammals inhabiting salt marshes might have acquired adaptations for ASL regulation. We studied the airways

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service