Skip to Content
Merck
All Photos(1)

Key Documents

MABS71

Sigma-Aldrich

Anti-phospho-MAPK 1/2 (Thr203/Ty205) Antibody, clone 14B9.1

clone 14B9.1, from mouse

Synonym(s):

Mitogen-activated protein kinase 3, MAP kinase 3, MAPK 3, ERT2, Extracellular signal-regulated kinase 1, ERK-1, Insulin-stimulated MAP2 kinase, MAP kinase isoform p44, p44-MAPK, Microtubule-associated protein 2 kinase, p44-ERK1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

14B9.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pThr203/pTyr205)

Gene Information

human ... MAPK3(5595)

General description

The extracellular signal-regulated kinases 1 and 2 (ERK1 and ERK2), also called p44 and p42 MAP kinases, are members of the Mitogen Activated Protein Kinase (MAPK) family of proteins found in all eukaryotes. The 44 kDa ERK1 and the 42 kDa ERK2 are highly homologous and both function in the same protein kinase cascade. For this reason, the two proteins are often referred to collectively as ERK1/2 or p44/p42 MAP kinase. The ERK1/2 signaling cascade has been shown to be a critical regulator of cell differentiation, cell physiology and neuronal function. Aberrant control of ERK1/2 activity has been implicated in a variety of pathological conditions, including cancer and autoimmune diseases, and efficient study methods are in demand. Extracellular signal/mitogen activated protein kinases (erk/MAPK) are a group of proline-directed serine/threonine kinases that are activated by dual phosphorylation of conserved threonine and tyrosine residues within a characteristic T X Y peptide motif. The mitogen-activated kinases erk1 (MAPK1) and erk2 (MAPK2) acquire full enzymatic activity upon phosphorylation of both threonine and tyrosine residues within the sequence motif T E Y.

Specificity

Antibody recognizes dual phosphorylation of MAPK1/2 at Thr203/Tyr204, but this antibody has not been used for the detection of single phosphorylation at either site.

Immunogen

KLH-conjugated linear peptide corresponding to human MAPK 1/2 phosphorylated at Thr203 and Ty205.

Application

Anti-phospho-MAPK 1/2 (Thr203/Ty205) Antibody, clone 14B9.1 is a highly specific mouse monoclonal antibody & that targets MAPK/Erk1/2 & has been tested in western blotting.
Research Category
Signaling
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: 0.1 µg/mL of this antibody detected phospho-MAPK 1/2 (Thr203/Ty205) in 200 µg of A431cell lysate and EGF treated A431 cell lysate.

Target description

~ 44 kDa observed

Linkage

Replaces: 05-481

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
A431cell lysate and EGF treated A431 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service