Skip to Content
Merck
All Photos(1)

Key Documents

MABN238

Sigma-Aldrich

Anti-Glucagon Antibody, clone 13D11.33

clone 13D11.33, from mouse

Synonym(s):

4030-01F, 4031-01F, AB932

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

13D11.33, monoclonal

species reactivity

human, rat

technique(s)

ELISA: suitable
immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
multiplexing: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GCG(2641)

General description

Glucagon is a hormone that is proteolytically cleaved into at least eight different functional peptides. It is produced by pancreatic cells and it has binding sites in the brain, intestines, and liver, among other tissues. Glucagon is secreted in response to hypoglycemia. Through the combined activation of Gsα- and Gq-coupled transmembrane receptors and their downstream signaling pathways, glucagon stimulates the release of glucose from the liver into the bloodstream. One important mechanism underlying this process is the activation of protein kinase A (PKA). PKA signals result in increased transcription of phosphoenolpyruvate carboxykinase, which encodes the rate-limiting enzyme in the production of glucose from noncarbohydrate substrates. PKA signals may also increase the breakdown of glycogen, and suppress glycolysis. Glucagon may also engage in calcium-dependent signaling via PLC. The importance of glucagon in regulating glucose homeostasis, in conjunction with insulin, is underscored by findings that glucagon and glucagon receptor signaling are disrupted in type II diabetes.

Immunogen

Ovalbumin-conjugated linear peptide corresponding to human Glucagon.

Application

Detect Glucagon using this Anti-Glucagon Antibody, clone 13D11.33 validated for use in IHC(P), Immunofluorescence, ELISA, Multiplexing.
Immunohistochemistry Analysis: A 1:8,000 dilution from a representative lot detected Glucagon in normal rat pancreas tissue (For enhanced staining results, it is recommended to use a 10,000 dilution of this antibody.).

ELISA Analysis: A representative lot detected Glucagon in ELISA.

Luminex® Analysis: A representative lot detected Glucagon in Luminex Assay.
Research Category
Neuroscience
Research Sub Category
Sensory & PNS

Quality

Evaluated by Immunofluorescence in normal rat pancreas tissue.

Immunofluorescence Analysis: A 1:8,000 dilution of this antibody detected Glucagon in normal rat pancreas tissue.

Target description

21 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Normal rat pancreas tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

Luminex is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Darian T Carroll et al.
Frontiers in endocrinology, 15, 1417437-1417437 (2024-08-08)
Using a non-human primate (NHP) model of maternal Western-style diet (mWSD) feeding during pregnancy and lactation, we previously reported altered offspring beta:alpha cell ratio in vivo and insulin hyper-secretion ex vivo. Mitochondria are known to maintain beta-cell function by producing
Jing Liu et al.
Developmental cell, 48(1), 49-63 (2019-01-09)
In the developing pancreas, transient Neurog3-expressing progenitors give rise to four major islet cell types: α, β, δ, and γ; when and how the Neurog3+ cells choose cell fate is unknown. Using single-cell RNA-seq, trajectory analysis, and combinatorial lineage tracing
Chandan Sona et al.
JCI insight, 7(6) (2022-02-09)
BACKGROUNDPathophysiology of type 1 diabetes (T1D) is illustrated by pancreatic islet infiltration of inflammatory lymphocytes, including CD8+ T cells; however, the molecular factors mediating their recruitment remain unknown. We hypothesized that single-cell RNA-sequencing (scRNA-Seq) analysis of immune cell populations isolated
Peter A Kropp et al.
American journal of physiology. Endocrinology and metabolism, 314(4), E308-E321 (2018-01-20)
The transcription factors pancreatic and duodenal homeobox 1 (Pdx1) and onecut1 (Oc1) are coexpressed in multipotent pancreatic progenitors (MPCs), but their expression patterns diverge in hormone-expressing cells, with Oc1 expression being extinguished in the endocrine lineage and Pdx1 being maintained
Karin J Bosma et al.
Molecular metabolism, 54, 101347-101347 (2021-10-10)
Type 2 diabetes is characterized by hyperglycemia and inflammation. Prostaglandin E2, which signals through four G protein-coupled receptors (EP1-4), is a mediator of inflammation and is upregulated in diabetes. We have shown previously that EP3 receptor blockade promotes β-cell proliferation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service