Skip to Content
Merck
All Photos(1)

Documents

MABC200

Sigma-Aldrich

Anti-APC Antibody, clone CC-1

clone CC-1, from mouse

Synonym(s):

Adenomatous polyposis coli protein, APC, Deleted in polyposis 2.5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

CC-1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... APC(324)

General description

Adenomatous polyposis coli protein (APC) is a ubiquitous multidomain protein that has a well documented role as a tumor suppressor. The mechanism of APC-mediated tumor suppression is still an area of active investigation; however, several studies suggest that APC is a negative regulator of the Wnt signaling pathway as it downregulates β-catenin via interactions with Axin/GSK3-β complex, thereby preventing transcription of genes such as MYC that contribute to cell proliferation. Defective APC proteins contribute to the initiation and proliferation of various types of cancers, including familial adenomatous polyposis. However, other studies have shown that APC interacts with a range of other proteins such as the EB1 microtubule-binding proteins, to regulate other processes such as cell migration.

Immunogen

Recombinant protein corresponding to human APC.

Application

Anti-APC Antibody, clone CC-1 detects levels of APC proteins & has been published & validated for use in IHC.
Immunohistochemistry Analysis: A representative lot from an independent laboratory detected APC in human brain tissue containing multiple schlerosis lesions (Saikali, P. et al. (2007). J Neurosci. 27(5):1220-1228.).

Immunohistochemistry Analysis: A representative lot from an independent laboratory detected APC in rat spinal cord injury tissue (McTique, D. M., et al. (2001). J Neurosci. 21(10):3392-3400.).

Quality

Immunohistochemistry Analysis: A 1:5 dilution from a representative lot detected APC in human colorectal cancer tissue, human smooth muscle tissue, and human colon tissue.

Target description

311 kDa calculated

Physical form

Format: Purified

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jasmin Steudler et al.
Glia, 70(11), 2045-2061 (2022-06-29)
Oligodendrocytes (ODCs) are myelinating cells of the central nervous system (CNS) supporting neuronal survival. Oxidants and mitochondrial dysfunction have been suggested as the main causes of ODC damage during neuroinflammation as observed in multiple sclerosis (MS). Nonetheless, the dynamics of
Emily G Baxi et al.
Glia, 62(9), 1513-1529 (2014-05-28)
Nerve conduction within the mammalian central nervous system is made efficient by oligodendrocyte-derived myelin. Historically, thyroid hormones have a well described role in regulating oligodendrocyte differentiation and myelination during development; however, it remains unclear which thyroid hormone receptors are required
Bastian G Brinkmann et al.
Neuron, 59(4), 581-595 (2008-09-02)
Understanding the control of myelin formation by oligodendrocytes is essential for treating demyelinating diseases. Neuregulin-1 (NRG1) type III, an EGF-like growth factor, is essential for myelination in the PNS. It is thus thought that NRG1/ErbB signaling also regulates CNS myelination
Qi-Yan Cai et al.
Anatomical record (Hoboken, N.J. : 2007), 292(4), 498-512 (2009-01-15)
The tumor suppressor phosphatase and tensin homologue (PTEN) is a protein and lipid phosphatase. PTEN mutations have been associated with a large number of human cancers. To understand the physiological role of PTEN in the brain and its relationship to
Kee-Yong Ha et al.
European spine journal : official publication of the European Spine Society, the European Spinal Deformity Society, and the European Section of the Cervical Spine Research Society, 17(6), 864-872 (2008-03-21)
The over-expression of excitotoxic neurotransmitter, such as glutamate, is an important mechanism of secondary injury after spinal cord injury. The authors examined the neuroprotective effect of pregabalin (GP) which is known as to reduce glutamate secretion, in a rat model

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service