Skip to Content
Merck
All Photos(1)

Documents

MAB948

Sigma-Aldrich

Anti-Coxsackievirus B3 Antibody, clone 280-5F-4E-5E

clone 280-5F-4E-5E, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

280-5F-4E-5E, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG2a

shipped in

wet ice

Specificity

Reacts with the Coxsackie virus B3. Neutralizes coxsackie virus B3 Nancy strain at a titer of 1:6,000.

Application

Anti-Coxsackievirus B3 Antibody, clone 280-5F-4E-5E is an antibody against Coxsackievirus B3 for use in IF.
IFA at 1:1,000. Dilute with buffer pH 7.5-8.0 containing protein or other stabilizing medium. Final working dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Format: Purified
Purified immunoglobulin. In 0.02M PB 0.25M NaCl, pH 7.6 with 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enteroviral infections in children with malignant disease: a 5-year study in a single institution.
Maria A Moschovi, Katerina Katsibardi, Maria Theodoridou, Athanassios G Michos et al.
The Journal of Infection null
Coxsackievirus B3 sequences in the myocardium of fatal cases in a cluster of acute myocarditis in Greece.
Spanakis, N; Manolis, EN; Tsakris, A; Tsiodras, S; Panagiotopoulos, T; Saroglou, G; Legakis, NJ
Journal of Clinical Pathology null
Yanhong Wei et al.
Molecules (Basel, Switzerland), 25(6) (2020-03-25)
Coxsackievirus B3 (CVB3) is the most common cause of acute and chronic viral myocarditis, primarily in children, while human adenovirus infections represent a significant cause of morbidity and mortality worldwide, in people of all ages. A series of novel 2-benzoxyl-phenylpyridine
Xiang Liu et al.
Scientific reports, 9(1), 10656-10656 (2019-07-25)
The Muc-1 oncoprotein is a tumor-associated mucin often overexpressed in pancreatic cancer. We report that knockout of Muc-1 reduced the degree of pancreatic inflammation that resulted from infection with Coxsackievirus B3 (CVB3) in a mouse model. CVB3-infected Muc-1-deficient (Muc-1KO) mice
Xiaowei Zhang et al.
Journal of virology, 87(22), 12407-12421 (2013-09-13)
Enteroviruses can frequently target the human central nervous system to induce a variety of neurological diseases. Although enteroviruses are highly cytolytic, emerging evidence has shown that these viruses can establish persistent infections both in vivo and in vitro. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service