Skip to Content
Merck
All Photos(1)

Key Documents

MAB88917

Sigma-Aldrich

Anti-Vitronectin Antibody, clone 8E6(LJ8)

clone 8E6(LJ8), Chemicon®, from mouse

Synonym(s):

Serum spreading factor, S-protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
8E6(LJ8), monoclonal
application:
ELISA
IHC
WB
species reactivity:
human
technique(s):
ELISA: suitable
immunohistochemistry: suitable
western blot: suitable
citations:
4

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

8E6(LJ8), monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... VTN(7448)

Specificity

The antibody reacts with human vitronectin, it does not react with human fibronectin (Hayman et al., 1983).

Immunogen

Purified human vitronectin from plasma.

Application

Anti-Vitronectin Antibody, clone 8E6(LJ8) detects level of Vitronectin & has been published & validated for use in ELISA, WB, IH.
Research Category
Cell Structure
Research Sub Category
ECM Proteins
Western blot: 5-20 μg/mL Stains cell-binding vitronectin.

Immunocytochemistry: 5-20 μg/mL

ELISA: 5-20 μg/mL

Optimal working dilutions must be determined by end user.

Linkage

Replaces: 04-1108

Physical form

Format: Purified
Purified immunoglobulin. Liquid. Buffer = 0.02M Phosphate buffer, 0.25M NaCl, pH 7.6 with 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 12 months.Avoid repeat freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takahiro Tougan et al.
Scientific reports, 8(1), 5052-5052 (2018-03-24)
The malaria parasite Plasmodium falciparum proliferates in the blood stream where the host immune system is most active. To escape from host immunity, P. falciparum has developed a number of evasion mechanisms. Serine repeat antigen 5 (SERA5) is a blood stage antigen
Heli Tiensuu et al.
BMC medicine, 20(1), 141-141 (2022-04-29)
Preterm birth is defined as live birth before 37 completed weeks of pregnancy, and it is a major problem worldwide. The molecular mechanisms that lead to onset of spontaneous preterm birth are incompletely understood. Prediction and evaluation of the risk
Cuihong Jia et al.
Experimental neurology, 323, 113088-113088 (2019-11-05)
Vitronectin (VTN) is a blood protein produced mainly by the liver. We show that VTN leaks from the bloodstream into the injury site and neighboring subventricular zone (SVZ) following ischemic stroke (middle cerebral artery occlusion, MCAO) in adult mice. MCAO
Quynh T Phan et al.
PLoS pathogens, 18(7), e1010681-e1010681 (2022-07-08)
During hematogenously disseminated candidiasis, blood borne fungi must invade the endothelial cells that line the blood vessels to infect the deep tissues. Although Candida albicans, which forms hyphae, readily invades endothelial cells, other medically important species of Candida are poorly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service