Skip to Content
Merck
All Photos(1)

Key Documents

MAB8257F

Sigma-Aldrich

Anti-Influenza A Antibody, nucleoprotein, clone A1, FITC-conjugated

clone A1, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

A1, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG2a

shipped in

wet ice

Specificity

Specific for the Influenza A nucleoprotein. Has stronger binding with N1-type Flu A. No cross reactivity seen to influenza B or other respiratory viruses.

Immunogen

Epitope: nucleoprotein
Influenza A

Application

Indirect Immunofluorescence

Optimal dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
This Anti-Influenza A Antibody, nucleoprotein, clone A1, FITC-conjugated is validated for use in IF for the detection of Influenza A.

Physical form

Purified, FITC conjugated. Liquid in 0.02M phosphate buffer, 0.25M sodium chloride, pH 7.6 with 15 mg/mL BSA and 0.1% sodium azide

Storage and Stability

Store at 2° to 8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheng-Chung Lee et al.
Scientific reports, 9(1), 4546-4546 (2019-03-16)
Influenza is a contagious acute respiratory disease caused by the influenza virus infection. Hemagglutinin (HA) is an important target in the therapeutic treatment and diagnostic detection of the influenza virus. Influenza A virus encompasses several different HA subtypes with different
Katherine S Xue et al.
eLife, 5, e13974-e13974 (2016-03-16)
RNA viruses rapidly diversify into quasispecies of related genotypes. This genetic diversity has long been known to facilitate adaptation, but recent studies have suggested that cooperation between variants might also increase population fitness. Here, we demonstrate strong cooperation between two
Kamonthip Rungrojcharoenkit et al.
PloS one, 15(8), e0237218-e0237218 (2020-08-08)
Influenza is an infectious respiratory illness caused by influenza viruses. Despite yearly updates, the efficacy of influenza vaccines is significantly curtailed by the virus antigenic drift and antigenic shift. These constant changes to the influenza virus make-up also challenge the
Surfactant protein A alters endosomal trafficking of influenza A virus in macrophages.
Yau, et al.
Frontiers in Immunology, 14, 919800-919800 (2023)
Bo Li et al.
Nature communications, 11(1), 164-164 (2020-01-11)
Host dependency factors that are required for influenza A virus infection may serve as therapeutic targets as the virus is less likely to bypass them under drug-mediated selection pressure. Previous attempts to identify host factors have produced largely divergent results

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service