Skip to Content
Merck
All Photos(1)

Key Documents

MAB5652

Sigma-Aldrich

Anti-Prox1 Antibody, clone 5G10

clone 5G10, Chemicon®, from mouse

Synonym(s):

Homeobox prospero-like protein PROX1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

5G10, monoclonal

species reactivity

human, mouse, chicken, lizard, rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Prox1(19130)

Application

This Anti-Prox1 Antibody, clone 5G10 is validated for use in WB, IH for the detection of Prox1.
Western Blot: 2 μg/mL
This antibody is recommended for immunohistochemistry use.

Optimal working dilutions must be determined by end user.

Target description

83 kDa

Physical form

Format: Purified

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marina Soulika et al.
Development (Cambridge, England), 143(11), 2012-2024 (2016-04-29)
Taste buds are sensory organs in jawed vertebrates, composed of distinct cell types that detect and transduce specific taste qualities. Taste bud cells differentiate from oropharyngeal epithelial progenitors, which are localized mainly in proximity to the forming organs. Despite recent
Henrik Boije et al.
Developmental biology, 330(1), 105-113 (2009-03-28)
We have addressed the question when horizontal cells in the chick retina are generated and undergo their terminal mitosis. Horizontal cell progenitors replicate their DNA early and migrate bi-directionally to the horizontal cell layer. It was hypothesized that the cells
Liyun Zhang et al.
PloS one, 8(2), e56108-e56108 (2013-02-14)
Proper retinal cell differentiation is essential for establishing a functional retina. The purpose of this study is to investigate the role of early growth response 1 (egr1), a transcription factor (TF) that has been reported to control eye development and
Linda T Doan et al.
PloS one, 7(9), e44009-e44009 (2012-09-18)
Insights into Bone morphogenetic protein (Bmp) functions during forebrain development have been limited by a lack of Bmp signaling readouts. Here we used a novel Bmp signaling reporter ("BRE-gal" mice) to study Bmp signaling in the dorsal telencephalon. At early
Pierluigi Valente et al.
Cerebral cortex (New York, N.Y. : 1991), 26(3), 1149-1167 (2015-01-13)
The functional consequence of γ-aminobutyric acid (GABA) release at mossy fiber terminals is still a debated topic. Here, we provide multiple evidence of GABA release in cultured autaptic hippocampal granule cells. In ∼50% of the excitatory autaptic neurons, GABA, VGAT

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service