Skip to Content
Merck
All Photos(1)

Key Documents

MAB380

Sigma-Aldrich

Anti-β-Tubulin III Antibody

CHEMICON®, mouse monoclonal, 5H1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

Product Name

Anti-Tubulin β Antibody, clone 5H1, ascites fluid, clone 5H1, Chemicon®

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

5H1, monoclonal

species reactivity

rat, human, bovine

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

isotype

IgM

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TTLL8(164714)

Specificity

Beta tubulin

Immunogen

Rat tubulin.

Application

Anti-Tubulin Antibody, β, clone 5H1 is an antibody against Tubulin for use in IH.
Immunohistochemistry at 1:300-1:1,000

Optimal working dilutions must be determined by end user.

IMMUNOHISTOCHEMISTRY PROTOCOL FOR MAB380

This antibody has been used successfully on 30 μm, free floating, 4% paraformaldehyde fixed rat brain tissue. All steps are performed under constant agitation. Suggested protocol follows.

1) 3 x 10 minute washes in TBS (with or without 0.25% Triton).

2) Incubate for 30 minutes in TBS with 3% serum (same as host from secondary antibody).

3) Incubate primary antibody diluted appropriately in TBS with 1% serum (same as host from secondary antibody) (with or without 0.25% Triton) for 2 hours at room temperature followed by 16 hours at 4°C.

4) 3 x 10 minute washes in TBS.

5) Incubate with secondary antibody diluted appropriately in TBS with1% serum (same as host from secondary antibody).

6) 3 x 10 minute washes in TBS.

7) ABC Elite (1:200 Vector Labs) in TBS.

8) 2 x 10 minute washes in TBS.

9) 1 x 10 minute wash in phosphate buffer (no saline).

10) DAB reaction with 0.06% NiCl added for intensification.

11) 2 x 10 minute washes in PBS.

12) 1 x 10 minute wash in phosphate buffer (no saline).
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Physical form

Ascites fluid containing sodium azide. Liquid

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to twelve months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human family with sequence similarity 60 member A (FAM60A) protein: a new subunit of the Sin3 deacetylase complex.
Smith, KT; Sardiu, ME; Martin-Brown, SA; Seidel, C; Mushegian, A; Egidy, R; Florens et al.
Molecular and Cellular Proteomics null
Carla J Cox et al.
Neurobiology of aging, 36(1), 178-187 (2014-10-16)
Flavonoids, a group of dietary polyphenols have been shown to possess cognitive health benefits. Epidemiologic evidence suggests that they could play a role in risk reduction in dementia. Amyloid precursor protein processing and the subsequent generation of amyloid beta (Aβ)
Kyle S McCommis et al.
Molecular metabolism, 5(8), 602-614 (2016-09-23)
Transport of pyruvate into the mitochondrial matrix by the Mitochondrial Pyruvate Carrier (MPC) is an important and rate-limiting step in its metabolism. In pancreatic β-cells, mitochondrial pyruvate metabolism is thought to be important for glucose sensing and glucose-stimulated insulin secretion.
Control of neural cell composition in poly(ethylene glycol) hydrogel culture with soluble factors.
Mooney, R; Haeger, S; Lawal, R; Mason, M; Shrestha, N; Laperle, A; Bjugstad, K; Mahoney, M
Tissue Engineering: Part A null
William E Barry et al.
eLife, 5 (2016-05-18)
Although mutations in HNF4A were identified as the cause of Maturity Onset Diabetes of the Young 1 (MODY1) two decades ago, the mechanisms by which this nuclear receptor regulates glucose homeostasis remain unclear. Here we report that loss of Drosophila

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service