Skip to Content
Merck
All Photos(1)

Key Documents

MAB3392

Sigma-Aldrich

Anti-Collagen Type III (COL3A1) Antibody

mouse monoclonal, 1E7-D7

Synonym(s):

collagen, type III, alpha 1, collagen, fetal, Ehlers-Danlos syndrome type IV, autosomal dominant, alpha1 (III) collagen, collagen alpha-1(III) chain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Collagen Type III Antibody, clone IE7-D7, clone 1E7-D7, from mouse

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1E7-D7, monoclonal

species reactivity

rat

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... COL3A1(1281)

General description

Type III collagen (also known as COL3A1), which adds structure and strength to connective tissues, is found in many places in the body, especially skin, lung, intestinal walls, and the walls of blood vessels. Collagen type III is initially produced as pro-collagen, a protein consisting of three pro-alpha1(III) chains that form the triple-stranded, rope-like molecule. After being synthesized, the pro-collagen molecule is modified by the cell. Enzymes modify the amino acids lysine and proline in the protein strands by adding chemical groups that are necessary for the strands to form a stable molecule and then later to crosslink to other molecules outside the cell. Other enzymes add sugars to the protein. The type III pro-collagen molecules are released from the cell and are processed by enzymes that clip small segments off either end of the molecules to form mature collagen. The mature collagen molecules assemble into fibrils. Cross-linking between molecules produces a very stable fibril, contributing to collagen′s tissue strengthening function.

Specificity

This antibody detects collagen type III. There is no evidence for cross reactivity with Collagen Types I, V and VI or connective tissue proteins (Elastin, Fibronectin and Laminin) at suggested working concentrations.

Immunogen

Epitope: N-terminus
Human type III collagen (Werkmeister, J.A., et al. 1990).

Application

ELISA Analysis: A previous lot of this antibody was used in ELISA (Werkmeister, J.A., et al., 1991).

Western Blot Analysis: A previous lot of this antibody was used to detect collagen type III in western blot under non-reduced conditions (Werkmeister J.A., et al., 1988; Ramshaw, J.S., et al., 1988).
Some Collagen samples can be contaminated with other Collagen Types. When purified Collagen is used in an application the purity of the Collagen sample should be verified by SDS-page to minimize the risk of false positives.

Immunohistochemistry Analysis: A previous lot of this antibody was used to detect collagen type III in immunohistochemistry (Werkmeister J.A., et al., 1989; Werkmeister J.A., et al., 1989; Werkmeister J.A., et al., 1988).
Research Category
Cell Structure
Research Sub Category
ECM Proteins
This Anti-Collagen Type III Antibody, clone IE7-D7 is validated for use in ELISA, WB, IH for the detection of Collagen Type III.

Quality

Evaluated by Immunohistochemistry in rat knee joint tissue.

Immunohistochemistry Analysis: A 1:600 dilution of this antibody detected Collagen Type III in rat knee joint tissue.

Target description

138 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat knee joint tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
This clone displays a high affinity for human, dog, rat, kangaroo and porcine Type III Collagens.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mingyu Cheng et al.
Tissue engineering. Part A, 16(5), 1479-1489 (2009-12-05)
Collagen-platelet (PL)-rich plasma composites have shown in vivo potential to stimulate anterior cruciate ligament (ACL) healing at early time points in large animal models. However, little is known about the cellular mechanisms by which the plasma component of these composites
Yvette M Coulson-Thomas et al.
Forensic science international, 251, 186-194 (2015-04-29)
Morphological and ultrastructural data from archaeological human bones are scarce, particularly data that have been correlated with information on the preservation of molecules such as DNA. Here we examine the bone structure of macroscopically well-preserved medieval human skeletons by transmission
Mark R Buckley et al.
Connective tissue research, 54(6), 374-379 (2013-10-04)
The mechanical properties of the human supraspinatus tendon (SST) are highly heterogeneous and may reflect an important adaptive response to its complex, multiaxial loading environment. However, these functional properties are associated with a location-dependent structure and composition that have not
Paolo Giannoni et al.
Biomedicines, 9(9) (2021-09-29)
Hypersensitivity pneumonitis (HP) is a diffuse interstitial lung disease (ILD) caused by the inhalation of a variety of antigens in susceptible individuals. Patients with fibrotic HP (fHP) may show histopathological and radiological manifestations similar to patients with idiopathic pulmonary fibrosis
Characterization of type I, III and V collagens in high-density cultured tenocytes by triple-immunofluorescence technique.
Gungormus, C; Kolankaya, D
Cytotechnology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service