Skip to Content
Merck
All Photos(1)

Key Documents

MAB3328

Sigma-Aldrich

Anti-MMP-14 Antibody, catalytic domain, clone LEM-2/15.8

clone LEM-2/15.8, Chemicon®, from mouse

Synonym(s):

MT1-MMP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

LEM-2/15.8, monoclonal

species reactivity

mouse, human

packaging

antibody small pack of 25 μg

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... MMP14(4323)

General description

MT1-MMP plays an important role during endothelial cell migration and matrix remodeling. Although the role of MT1-MMP in endothelial cell motility is not fully characterized, its activity appears to modulate endothelial migration, invasion, and formation of capillary tubes during the angiogenic response (Galvez, 2001). Mt1-MMP also appears to play a key role in monocyte revruitment during inflammation (Salomon, 2005).

Specificity

LEM-2/15.8 reacts with human MT1-MMP and displays crossreactivity with mouse specimens. This antibody was generated against the catalytic domain of MT1-MMP and is able to inhibit enzyme activity.

Immunogen

Epitope: catalytic domain
Synthetic peptide: amino acid sequence 218-233 within the catalytic domain

Application

Detect MMP-14 using this Anti-MMP-14 Antibody, catalytic domain, clone LEM-2/15.8 validated for use in ELISA, IP & WB.
Research Category
Cell Structure
Research Sub Category
MMPs & TIMPs
Western Blot

Immunohistochemistry: Frozen and paraffin-embedded tissues

Immunofluorescence

Flow Cytometry

Blocking: 10-15μg/mL

Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified
Purified immunoglobulin by Protein A chromatography . Liquid in 0.2M phosphate, 0.25M NaCl, pH 7.6, containing 0.1% sodium azide.

Storage and Stability

Maintain at 2° to 8°C for up to 12 months from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J K Björk et al.
Oncogene, 35(14), 1770-1784 (2015-06-30)
Heat-shock factors (HSFs) are key transcriptional regulators in cell survival. Although HSF1 has been identified as a driver of carcinogenesis, HSF2 has not been explored in malignancies. Here, we report that HSF2 suppresses tumor invasion of prostate cancer (PrCa). In
Charles Marusak et al.
Pharmacological research, 113(Pt A), 515-520 (2016-10-21)
MT1-MMP and MMP2 have been implicated as pro-tumorigenic and pro-metastatic factors in a wide variety of cancers including melanoma. We have previously demonstrated that MT1-MMP is highly expressed in melanoma where it promotes melanoma cell invasion and metastasis in part
Susan H Taylor et al.
eLife, 4, e09345-e09345 (2015-09-22)
Type I collagen-containing fibrils are major structural components of the extracellular matrix of vertebrate tissues, especially tendon, but how they are formed is not fully understood. MMP14 is a potent pericellular collagenase that can cleave type I collagen in vitro.
G Daniel Grass et al.
Journal of cell science, 125(Pt 3), 777-788 (2012-03-06)
A defining feature of malignant tumor progression is cellular penetration through the basement membrane and interstitial matrices that separate various cellular compartments. Accumulating evidence supports the notion that invasive cells employ specialized structures termed invadopodia to breach these structural barriers.
Karina Di Gregoli et al.
Arteriosclerosis, thrombosis, and vascular biology, 34(9), 1990-2000 (2014-07-06)
Our recent studies have highlighted membrane type-1 matrix metalloproteinase (MMP)-14 as a selective marker for an invasive subset of macrophages potentially related to atherosclerotic plaque progression. Moreover, colony stimulating factors (CSF) may exert divergent effects on macrophage MMP expression, possibly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service