Skip to Content
Merck
All Photos(1)

Key Documents

MAB3228

Sigma-Aldrich

Anti-Cytokeratin 5/8

Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

monoclonal

species reactivity

rat, mouse, rabbit, pig, canine, hamster, human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... KRT5(3852)

General description

Keratins are the intermediate filament proteins of epithelia that display high degree of molecular diversity. They are heteropolymeric filaments formed by pairing of type I and type II keratins that are expressed in a highly specific patterns depending on the epithelial type and stage of cellular differentiation. Keratins contain a central rod domain of about 310 amino acids with alpha-helical conformation bordered by non-helical head and tail domains of variable length. The head domain consists of subdomains V1 and H1. The central alpha helical rod domain is composed of subdomains 1A, 1B, 2A, and 2B connected by the linkers L1, L12, and L2. The tail domain then consists of subdomains H2 and V2. They provide stability between epithelial cells and their attachment to basement membrane. An important unique feature of cytokeratins is their epithelial cell-type-specific expression. Hence, they can be used as tumor markers and epithelial differentiation markers. Cytokeratin-5 (CK5) is a type II cytoskeletal protein that is primarily expressed in basal keratinocytes in the epidermis, specifically in the stratified epithelium lining the skin and digestive tract. It can serve as a biomarker for several different types of cancer, including breast and lung cancers. Cytokeratin-8 (CK8) is a type II, neutral to basic protein that together with cytokeratin-19 (KRT19) helps to link the contractile apparatus to dystrophin at the costameres of striated muscle. It can undergo phosphorylation on three major serine residues: Serine 23, 431, and 73. Serine 23 is shown to be highly conserved in all type II keratins. It can also undergo O-glycosylation in a cell cycle-dependent manner and glycosylation increases its solubility and reduces stability by inducing proteasomal degradation. CK8 expression has been correlated with malignancy in leukoplakia and carcinomas of the head and neck and its expression has been observed in all non-small-cell lung cancers.

Specificity

Cytokeratin 5 and 8. MAB3228 is a broadly reacting cytokeratin antibody specific for cytokeratin 5 and cytokeratin 8. This antibody recognizes virtually all epithelial tissues and carcinomas.

Immunogen

Cytokeratins from the human lung cancer cell line MR21.

Application

Research Category
Cell Structure
Research Sub Category
Cytokeratins
This Anti-Cytokeratin 5/8 is validated for use in FC, WB, IC, IH(P) for the detection of Cytokeratin 5.
Western blot

Immunohistochemistry on frozen and paraffin embedded tissue sections.

Immunocytochemistry

Flow cytometry

Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified
Liquid in buffer with 0.1% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots up to 6 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Homeodomain interacting protein kinase 2 regulates postnatal development of enteric dopaminergic neurons and glia via BMP signaling.
Chalazonitis, A; Tang, AA; Shang, Y; Pham, TD; Hsieh, I; Setlik, W; Gershon, MD; Huang, EJ
The Journal of Neuroscience null
Masakazu Inada et al.
International journal of molecular medicine, 37(6), 1521-1527 (2016-04-29)
The 293 cell line, used extensively in various types of studies due to the ease with which these cells can be transfected, was thought to be derived by the transformation of primary cultures of human embryonic kidney cells with sheared adenovirus
Amy P Wong et al.
American journal of physiology. Lung cellular and molecular physiology, 293(3), L740-L752 (2007-07-10)
It has been suggested that some adult bone marrow cells (BMC) can localize to the lung and develop tissue-specific characteristics including those of pulmonary epithelial cells. Here, we show that the combination of mild airway injury (naphthalene-induced) as a conditioning
Hsing-Hui Wang et al.
The Prostate, 75(14), 1620-1631 (2015-07-16)
The presence of inflammation in prostate cancer (PCa) and benign prostate hyperplasia (BPH) has been well described but the cellular mechanisms by which inflammation modulates the prostate are currently unclear. Prostate stem cells (PSC) not only maintain prostate homeostasis but
Nesrine Benkafadar et al.
EMBO molecular medicine, 9(1), 7-26 (2016-10-30)
Cisplatin is a widely used chemotherapy drug, despite its significant ototoxic side effects. To date, the mechanism of cisplatin-induced ototoxicity remains unclear, and hearing preservation during cisplatin-based chemotherapy in patients is lacking. We found activation of the ATM-Chk2-p53 pathway to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service