Skip to Content
Merck
All Photos(1)

Key Documents

MAB3083

Sigma-Aldrich

Anti-Calpain Antibody, small subunit of µ- or m-Calpains (Calpain I or II), clone P1

ascites fluid, clone P1, Chemicon®

Synonym(s):

Anti-CALPAIN4, Anti-CANP, Anti-CANPS, Anti-CAPN4, Anti-CDPS, Anti-CSS1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

P1, monoclonal

species reactivity

human, bovine

should not react with

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CAPNS1(826)

General description

Calpains are non-lysosomal cysteine proteeases which absolutely require calcium for activity. There are two major calpain isozymes: μ-calpain and m-calpain, which require micromolar or millimolar calicum for activity, respectively, in invitro assays. The major calpain isozymes are composed of two subunits: an 80kDa catalytic subunit; and a 30kDa regulatory subunit. The large and small subunits remain associated in the presence of calcium after proteolytic activation of the zymogen.

Specificity

Reacts with the small subunit of human erthrocyte μ-calpain (Calpain I) or bovine myocardial m-calpain (Calpain II).

Immunogen

Bovine myocardial m-calpain.
Epitope: small subunit of μ- or m-Calpains (Calpain I or II)

Application

Immunoblot: 1:500-1,000. The small subunit of either μ-calpain m-calpain is 30kDa. ELISA: 1:50,000 (against the immunogen). Immunohistochemistry: 1:100 on 4% paraformaldehyde or 1% paraformaldehyde/4% glutaraldehyde fixed tissue. Immunocytochemistry: 1:100. Optimal working dilutions must be determined by the end user.
This Anti-Calpain Antibody, small subunit of μ-or m-Calpains (Calpain I or II), clone P1 is validated for use in ELISA, WB, IC, IH for the detection of Calpain.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Critical role for cathepsin B in mediating caspase-1-dependent interleukin-18 maturation and caspase-1-independent necrosis triggered by the microbial toxin nigericin.
Hentze, H; Lin, XY; Choi, MS; Porter, AG
Cell Death and Differentiation null
Hyun-Hwa Cha et al.
Reproductive sciences (Thousand Oaks, Calif.), 21(5), 612-622 (2013-11-08)
To investigate the interconnection of apoptosis and autophagy in trophoblastic cells, we treated JEG-3 cells with tumor necrosis factor α (TNF-α) after transfecting LC3 or Beclin 1 or calpain small interfering RNA (siRNA), which blocks cleavage of autophagy-related gene 5
A radioimmunologic technique for assessing calpain activation in cells.
R L Mellgren
Methods in molecular biology (Clifton, N.J.), 144, 161-169 (2000-05-20)
Kathrin Stelzner et al.
mBio, 11(6) (2020-12-17)
The opportunistic human pathogen Staphylococcus aureus causes serious infectious diseases that range from superficial skin and soft tissue infections to necrotizing pneumonia and sepsis. While classically regarded as an extracellular pathogen, S. aureus is able to invade and survive within
Cheng-Wei Lu et al.
Biomedicines, 9(3) (2021-04-04)
Cognitive impairment is not only associated with seizures but also reported as an adverse effect of antiepileptic drugs. Thus, new molecules that can ameliorate seizures and maintain satisfactory cognitive function should be developed. The antiepileptic potential of asiatic acid, a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service