Skip to Content
Merck
All Photos(1)

Key Documents

MAB2077Z

Sigma-Aldrich

Anti-Integrin αVβ6 Antibody, clone 10D5, azide free

clone 10D5, Chemicon®, from mouse

Synonym(s):

MAB2077

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10D5, monoclonal

species reactivity

human, rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunoprecipitation (IP): suitable

isotype

IgG2a

suitability

not suitable for Western blot
not suitable for immunohistochemistry

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGAV(3685)

General description

The integrin family of cell adhesion receptors consists of at least 19 membrane-associated heterodimers, composed of an alpha and beta subunit that associate in a non-covalent manner. To date, 16 different alpha-subunits and 8 different beta-subunits have been identified. The structure and functional diversity of the integrin family are based upon the pairing abilities of the individual alpha and beta subunits. Key to these molecular interactions between the integrin receptors and their respective ligands is the recognition of the Arg-Gly-Asp (RGD) sequence, known to be present in the extracellular matrix components fibronectin, vitronectin, collagen, fibrinogen, and von Willebrand factor (Cheresh, 1991). The involvement of integrins in vascular proliferation, adhesion, and wound repair has been well documented. Integrin alphaV is a type I membrane glycoptroein with no I (inserted) domain. The alphaV protein is post-translationally cleaved into a heavy and light chain, which is expressed on the cell surface as a disulfide linked polypeptide.

The integrin alphavbeta6 is expressed on a variety of epithelial cells during dynamic processes including organogenesis, tissue injury and malignant transformation.

Specificity

Monoclonal antibody MAB2077Z, raised by immunizing beta6 knockout mice with wild type murine keratinocytes, recognizes human and murine alphaVbeta6 by flow cytometry and immunoprecipitation and has blocking activity against murine and human alphaVbeta6. 100% inhibition of alphaVbeta6-mediated cell adhesion and cell migration are achieved at a concentration of <10 μg/mL. Because the antibody was raised in knockout mice, it is likely to recognize the receptor in other species, however it has been evaluated only in human and mouse.The antibody is specific for alphaVbeta6 and does not recognize aV or other aV integrins.

Immunogen

Immunization of beta 6 knockout mice with wild type murine keratinocytes.

Application

Anti-Integrin αVβ6 Antibody, clone 10D5, azide free is an antibody against Integrin αVβ6 for use in FC & IP.
Flow cytometry: Effective for mouse and human.

Immunoprecipitation: Effective; however because this antibody is an IgG2a, protein G or rabbit anti-mouse IgG is helpful for capture

Function blocking: 100% inhibition of alphaVbeta6-mediated cell adhesion and cell migration are achieved at a concentration of 100 μg/mL.

Not effective for western blot or immunohistochemistry.

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Integrins

Physical form

Format: Purified
Protein A Purified mouse immunoglobulin in 20 mM sodium phosphate, 250 mM NaCl, pH. 7.6, with no preservatives.
Protein A purified

Storage and Stability

Maintain for 1 year at 2–8°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
HT-29 colon carcinoma cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicholas Juleff et al.
PloS one, 3(10), e3434-e3434 (2008-10-23)
Foot-and-mouth disease virus (FMDV) is one of the most contagious viruses of animals and is recognised as the most important constraint to international trade in animals and animal products. Two fundamental problems remain to be understood before more effective control
Chemically programmed antibodies targeting multiple alpha(v) integrins and their effects on tumor-related functions in vitro.
Goswami, RK; Bajjuri, KM; Forsyth, JS; Das, S; Hassenpflug, W; Huang, ZZ; Lerner et al.
Bioconjugate Chemistry null
A peptide selected by biopanning identifies the integrin alphavbeta6 as a prognostic biomarker for nonsmall cell lung cancer.
Elayadi, AN; Samli, KN; Prudkin, L; Liu, YH; Bian, A; Xie, XJ; Wistuba, II; Roth et al.
Cancer Research null
The ability of integrin alpha(v)beta(3) To function as a receptor for foot-and-mouth disease virus is not dependent on the presence of complete subunit cytoplasmic domains.
Neff, S; Baxt, B
Journal of virology null
Claire S Reader et al.
The Journal of pathology, 249(3), 332-342 (2019-07-02)
Pancreatic ductal adenocarcinoma (PDAC) has a 5-year survival rate of less than 4% and desperately needs novel effective therapeutics. Integrin αvβ6 has been linked with poor prognosis in cancer but its potential as a target in PDAC remains unclear. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service