Skip to Content
Merck
All Photos(1)

Key Documents

MAB1932

Sigma-Aldrich

Anti-Fibronectin Antibody

CHEMICON®, mouse monoclonal, 875A51

Synonym(s):

Anti-CIG, Anti-ED-B, Anti-FINC, Anti-FN, Anti-FNZ, Anti-GFND, Anti-GFND2, Anti-LETS, Anti-MSF, Anti-SMDCF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

product name

Anti-Fibronectin Antibody, mid-molecule, clone 875A51, clone 875A51, Chemicon®, from mouse

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

875A51, monoclonal

species reactivity

canine, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

Gene Information

human ... FN1(2335)

Specificity

Reacts with the middle (central domain) of the human plasma fibronectin monomer but does not interfere with cell adhesion. Positive reactivity with human and canine samples. Negative reactivity with chick.

Immunogen

Epitope: mid-molecule
Human plasma fibronectin.

Application

Detect Fibronectin using this Anti-Fibronectin Antibody, mid-molecule, clone 875A51 validated for use in ELISA & WB.
ELISA

Microscopy

Western blotting at 1-5 μg/ml

Final working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Physical form

Format: Purified
Liquid in 0.02M PB with 0.25M NaCl,pH=7.6, 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hee Doo Lee et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 26(7), 3084-3095 (2012-04-17)
A disintegrin and metalloproteinase 15 (ADAM15), the only ADAM protein containing an Arg-Gly-Asp (RGD) motif in its disintegrin-like domain, is a widely expressed membrane protein that is involved in tumor progression and suppression. However, the underlying mechanism of ADAM15-mediated tumor
Ursula Hartmann et al.
Journal of neurochemistry, 125(3), 399-409 (2013-02-20)
The testicans are a three-member family of secreted proteoglycans structurally related to the BM-40/secreted protein acidic and rich in cystein (SPARC) osteonectin family of extracellular calcium-binding proteins. In vitro studies have indicated that testicans are involved in the regulation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service