Skip to Content
Merck
All Photos(1)

Key Documents

MAB1628

Sigma-Aldrich

Anti-Myosin Antibody, slow muscle, clone NOQ7.5.4D

clone NOQ7.5.4D, Chemicon®, from mouse

Synonym(s):

Anti-CMD1S, Anti-CMH1, Anti-MPD1, Anti-MYHCB, Anti-SPMD, Anti-SPMM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

NOQ7.5.4D, monoclonal

species reactivity

rat, feline, human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
radioimmunoassay: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MYH7B(57644)

Specificity

Slow myosin heavy chain. Clearly identifies Type 1 fibers. Within skeletal muscle MAB1628 is specific for slow myosin heavy chain in a wide variety of species. It reacts strongly with rat and feline slow myosin heavy chain. MAB1628 also identifies beta (slow) myosin heavy chain in heart ventricles.

Immunogen

Epitope: slow muscle
Myosin purified from myofibrils isolated from histochemically mixed human skeletal muscle.

Application

Anti-Myosin Antibody, slow muscle, clone NOQ7.5.4D is an antibody against Myosin for use in RIA, WB, IH.
Immunohistochemistry: frozen and formalin fixed sections.

Immunoblotting

RIA

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jenny Lund et al.
Scientific reports, 8(1), 9814-9814 (2018-07-01)
Once assumed only to be a waste product of anaerobe glycolytic activity, lactate is now recognized as an energy source in skeletal muscles. While lactate metabolism has been extensively studied in vivo, underlying cellular processes are poorly described. This study
Camila Silva Foresto et al.
Journal of applied physiology (Bethesda, Md. : 1985), 121(3), 646-660 (2016-07-23)
Muscle loss occurs following injury and immobilization in adulthood and childhood, which impairs the rehabilitation process; however, far fewer studies have been conducted analyzing atrophic response in infants. This work investigated first the morphological and molecular mechanisms involved in immobilization-induced
Lipid in skeletal muscle myotubes is associated to the donors' insulin sensitivity and physical activity phenotypes.
Bajpeyi, S; Myrland, CK; Covington, JD; Obanda, D; Cefalu, WT; Smith, SR; Rustan, AC; Ravussin, E
Obesity (Silver Spring, Md.) null
Jenny Lund et al.
PloS one, 12(4), e0175441-e0175441 (2017-04-14)
Physical activity has preventive as well as therapeutic benefits for overweight subjects. In this study we aimed to examine effects of in vivo exercise on in vitro metabolic adaptations by studying energy metabolism in cultured myotubes isolated from biopsies taken
Christian M Girgis et al.
Journal of cachexia, sarcopenia and muscle, 10(6), 1228-1240 (2019-06-22)
It has long been recognized that vitamin D deficiency is associated with muscle weakness and falls. Vitamin D receptor (VDR) is present at very low levels in normal muscle. Whether vitamin D plays a direct role in muscle function is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service