Skip to Content
Merck
All Photos(1)

Key Documents

MAB1012

Sigma-Aldrich

Anti-Alkaline Phosphatase Antibody, E. coli, bacterial only

ascites fluid, Chemicon®

Synonym(s):

AP, Alk. Phos.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

monoclonal

species reactivity

E. coli

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

Escherichia coli ... PhoA(945041)

Specificity

Alkaline phosphatase (AP). MAB1012 has a high affinity and recognizes an AP determinant resistant to denaturation by SDS-PAGE. It is therefore ideally suited for sensitive and specific detection of AP fusion proteins by Western blot analysis of E. coli transformants expressing fusion products.

Immunogen

Epitope: E. coli, bacterial only
F201C1B

Application

Anti-Alkaline Phosphatase Antibody, E. coli, bacterial only detects level of Alkaline Phosphatase & has been published & validated for use in IP, WB & IC.
Western blot at 1:5,000. 42-45kDa on SDS-PAGE,reducing gels for natural E. coli alkaline phosphatase monomer. Alkaline phosphatase fusion proteins will vary depending upon the target fusion protein.

Immunocytochemistry: reacts with E.coli. AP fusion protein targets in acetone fixed cell preparations. 1:4000, other fixatives or conditions untested.

ASSAY:

Preparation of E. coli TnphoA transformants: E. coli strain CC118 was transformed with plasmid pGEM-3Z containing TnphoA insertional mutations in the p101 gene of Mycoplasma hyorhinis, which encodes a protein with a typical N-terminal prokaryotic single peptide (Yogev et al. 1991).

Identification of fusion protein with MAB1012 transformants: Transformants are grown in 2XYT medium to OD600=0.6. Cells were centrifuged 3 minutes at 10,000 x g, suspended in SDS-PAGE sample buffer, heated at 100°C for 5 minutes, frozen and thawed and centrifuged as above at room temperature to remove insoluble material. The sample is applied at 9% to a SDS-PAGE gel, and Western immunoblot is performed as described (Yogev et al. 1991).

Immunoprecipitation: 5μL of antibody per 500μL of lysate in RIPA or 0.5% triton X-100 solutions.

Optimal working dilutions must be determined by end user.

Physical form

Ascites. Contains no preservative.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christina Grogan et al.
Frontiers in microbiology, 12, 633667-633667 (2021-03-06)
Mosquitoes vector many pathogens that cause human disease, such as malaria that is caused by parasites in the genus Plasmodium. Current strategies to control vector-transmitted diseases are hindered by mosquito and pathogen resistance, so research has turned to altering the
May N Taw et al.
ACS synthetic biology, 10(11), 2947-2958 (2021-11-11)
Escherichia coli remains one of the preferred hosts for biotechnological protein production due to its robust growth in culture and ease of genetic manipulation. It is often desirable to export recombinant proteins into the periplasmic space for reasons related to
Lisa R Knoke et al.
Redox biology, 64, 102800-102800 (2023-07-07)
The thiol redox balance in the periplasm of E. coli depends on the DsbA/B pair for oxidative power and the DsbC/D system as its complement for isomerization of non-native disulfides. While the standard redox potentials of those systems are known
Tobias Schilling et al.
ACS synthetic biology, 12(12), 3656-3668 (2023-11-27)
Bacillus subtilis is a major workhorse for enzyme production in industrially relevant quantities. Compared to mammalian-based expression systems, B. subtilis presents intrinsic advantages, such as high growth rates, high space-time yield, unique protein secretion capabilities, and low maintenance costs. However
Functional and topological analysis of the Burkholderia cenocepacia priming glucosyltransferase BceB, involved in the biosynthesis of the cepacian exopolysaccharide.
Videira, PA; Garcia, AP; Sa-Correia, I
Journal of Bacteriology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service