Skip to Content
Merck
All Photos(1)

Key Documents

KX0020

Supelco

Kerosene

GR

Synonym(s):

Kerosene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352401
EC Index Number:
265-150-3
bp:
180-188 °C
vapor pressure:
0.75 hPa ( 20 °C)

vapor pressure

0.75 hPa ( 20 °C)

Quality Level

form

liquid

impurities

≤1 ppm Sulfur (S)

color

APHA: ≤20

bp

180-188 °C

transition temp

flash point 54 °C

shipped in

ambient

storage temp.

room temp

Application

GR
Odorless solvent.

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Analysis Note

Boiling range171-191°CColor (APHA)20 maxFormClear odorless liquidInfrared SpectrumConforms to standardSpecific Gravity at 15.6/15.6°C0.756-0.761Sulfur (S)1ppm max
Evaporation Number: 0.16

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Carc. 1B - Flam. Liq. 3 - Muta. 1B - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

100.4 - 125.6 °F

Flash Point(C)

38 - 52 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service