JH-10L-BK
Intermediates for Further Manufacturing Use (FFMU) - ABO Specificities
Anti-A from the clone BIRMA-1, Murine IgM, Intermediate for further manufacturing use
Synonym(s):
Anti-A (clone BIRMA-1), FFMU
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
unpurified
clone
BIRMA-1, monoclonal
form
liquid
packaging
pack of 10 L
concentration
1-11 mg/mL protein (Total Protein Range)
technique(s)
agglutination assay: suitable
pH
6.7-7.7
isotype
IgM
shipped in
ambient
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
For the formulation of Anti-A or Anti-A+B blood typing reagents or devices.
The BIRMA-1 antibody reacts strongly with all common forms of the A antigen, including A2B. When used at appropriate concentrations it will directly agglutinate most examples of A<sub>x</sub>. It has the benefit of not agglutinating red cells classified as having the B(A) phenomenon, nor of agglutinating unmodified or enzyme treated B or O red cells. It is a proven raw material for the manufature of Anti-A reagents.
Contains 0.1% (w/v) Sodium Azide; Shelf Life: 24 months from date of filtration.
Licensed by the US FDA under a shared manufacturing agreement.
Application
Legal Information
Disclaimer
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service