Skip to Content
Merck
All Photos(1)

Key Documents

GR01

Sigma-Aldrich

Anti-EGFR (Ab-1) Mouse mAb (528)

liquid, clone 528, Calbiochem®

Synonym(s):

Anti-Epidermal Growth Factor Receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

528, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

should not react with

mouse, rat

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

isotype

IgG2a

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... EGFR(1956)
mouse ... Egfr(13649)
rat ... Egf(25313)

General description

Anti-EGFR (Ab-1), mouse monoclonal, clone 528, recognizes EGF receptor protein in A431 cells and normal skin. It is validated for use in ELISA, IF, IP, IHC (frozen,paraffin) & neutralization studies.
Purified mouse monoclonal antibody generated by immunizing BALB/c mice with the specified immunogen and fusing splenocytes with NS-1 mouse myeloma cells (see application references. Recognizes the ~170 kDa EGF receptor.
Recognizes the ~170 kDa EGF receptor protein in A431 cells and normal skin.

Application

Flow Cytometry (5 µg/ml)

Frozen Sections (2.5 µg/ml)

Immunofluorescence (5 µg/ml)

Immunoprecipitation(1 µg/sample)

Neutralization Studies (see comments and application references)

Paraffin Sections (see application references)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Analysis Note

Negative Control
HL-60 cells or human brain
Positive Control
A-431 or normal skin tissue

Other Notes

Hunter T. 1984. Nature311, 414.
Masui, H., et al. 1984. Cancer Res.44, 1002.
Reynolds, F. H., Jr., et al. 1981. Nature 292, 259.
Ushiro, H. and Cohen, S. 1980. J. Biol. Chem.255, 8363.
This antibody inhibits EGF binding to its receptor and is an antagonist of in vivo EGF-stimulated tyrosine kinase activity (see application references). Also inhibits A431 tumor formation in nude mice. For neutralization studies, use Cat. No. GR01L; Cat. No. GR01 contains azide, which is toxic to cells. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rozanne Lee et al.
Journal of biomolecular screening, 13(3), 210-217 (2008-03-04)
Monoclonal antibodies (mAb) are not only useful reagents but also represent a promising type of therapeutics due to their high affinity and exquisite specificity for their antigens. A critical step in mAb generation is to identify antigen-specific antibodies. Although enzyme-linked
Inmaculada Bañón-Rodríguez et al.
The EMBO journal, 33(2), 129-145 (2014-01-15)
Establishing the correct orientation of the mitotic spindle is an essential step in epithelial cell division in order to ensure that epithelial tubules form correctly during organ development and regeneration. While recent findings have identified some of the molecular mechanisms
David Cachia et al.
Journal of neuro-oncology, 125(2), 401-410 (2015-09-12)
Gliosarcoma is classified by the World Health Organization as a variant of glioblastoma. These tumors exhibit biphasic histologic and immunophenotypic features, reflecting both glial and mesenchymal differentiation. Gliosarcomas can be further classified into primary (de novo) tumors, and secondary gliosarcomas
F Terzi et al.
The Journal of clinical investigation, 106(2), 225-234 (2000-07-21)
The role of EGF in the evolution of renal lesions after injury is still controversial. To determine whether the EGF expression is beneficial or detrimental, we generated transgenic mice expressing a COOH-terminal-truncated EGF-R under the control of the kidney-specific type
Valencio Salema et al.
mAbs, 8(7), 1286-1301 (2016-07-30)
Most therapeutic antibodies (Abs) target cell surface proteins on tumor and immune cells. Cloning of Ab gene libraries in E. coli and their display on bacteriophages is commonly used to select novel therapeutic Abs binding target antigens, either purified or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service