GF439-M
HumanKine® TGF beta 1, Human Recombinant Xeno-Free
Synonym(s):
TGFB, TGFb1, Transforming Growth Factor-β1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.75
Recommended Products
General description
HumanKine growth factors are expressed in a proprietary human 293 protein expression system and are manufactured without the use of any animal products and are considered Xeno-Free.
Transforming growth factor beta (TGF-B) is a multifunctional cytokine that controls proliferation, differentiation, and other functions in many cell types. TGF-B acts synergistically with TGF-A in inducing transformation. It also acts as a negative autocrine growth factor. TGF-B regulates the actions of many other growth factors and determines a positive or negative direction of their effects. It plays an important role in bone remodeling. It is a potent stimulator of osteoblastic bone formation, causing chemotaxis, proliferation and differentiation in committed osteoblasts.
Application
Research Category
Stem Cell Research
Stem Cell Research
Research Sub Category
Growth Factors & Receptors
Growth Factors & Receptors
Quality
Bioactivity assay: Determined by the dose dependent inhibition of IL-4 induced proliferation of mouse HT-2 cells (BALB/c spleen activated by sheep erythrocytes in the presence of IL-2). ED50 is typically 0.1-0.5 ng/ml.
Physical form
Product is presented in 50mM NaOAc pH 4.5 and filtered through a 0.2 micron filter before lyophilization.
Preparation Note
HumanKine TGF beta1 is manufactured using all non-animal reagents and is considered Xeno-Free
Storage and Stability
Store at -20°C for up to 4 months from date of receipt. Further dilutions should be made into appropriate buffer or medium to which protein has been added.
Legal Information
HumanKine is a registered trademark of Proteintech Group, Inc. and Humanzyme, Inc
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service