EZFITL0W01
EZ-Fit® Manifold,1-place for Microfil® V or Microfil® S filtration units
General microbiological analysis
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116129
NACRES:
NB.24
Recommended Products
material
stainless steel
Sterilization Compatibility
autoclavable compatible
manufacturer/tradename
EZ-Fit®
input
sample type pharmaceutical(s)
sample type cosmetics
sample type beer
sample type bottled water
application(s)
bioburden testing
cosmetics
environmental
food and beverages
water monitoring
shipped in
ambient
Related Categories
General description
EZ-Fit Manifold is available with the new Microfil head for use with presterilized, ready-to-use funnels and membranes.
Application
General microbiological analysis
Legal Information
EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service