Skip to Content
Merck
All Photos(1)

Key Documents

EP12

Millipore

NovaSeptum® GO In-line Holder, 1-port

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104105

material

303 stainless steel nut (Locking nut)
polyester ring(s) (safety ring)

Quality Level

product line

NovaSeptum® GO

feature

autoclavable

parameter

-80-150 °C temp. range (-112-302 °F)
6 bar max. pressure (87 psi)

application(s)

cell therapy
gene therapy
life science and biopharma
mAb
ophthalmics
parenterals
pathogen testing
pharma/biopharma processes
pyrogen testing
sterile sampling
sterility testing
viral therapy

storage temp.

room temp

General description

Device configuration: Sterile Sampling connectors
The NovaSeptum® GO holder for In-line configuration is designed to be used in combination with NovaSeptum® GO sampling units and is designed either to be welded on a pipe, either for installation on a pipe with Tri-calmp connectors. Electropolished and glass-blasted

Application

Sterility testing, pyrogenic testing, endotoxin testing, chemical analysis, pH analysis, and fermentation applications

Packaging

EP12/127x94-3D5 (1 unit per box)
EP12/191x158-3D5 (1 unit per box)
EP12/130x100-3D5 (1 unit per box)
EP12/190x160-3D5 (1 unit per box)
EP12/127x94-3D4 (1 unit per box)
EP12/191x158-3D4 (1 unit per box)
EP12/130x100-3D4 (1 unit per box)
EP12/190x160-3D4 (1 unit per box

Components

  • Magazine: Polyphenylene sulfide (PPS)
  • Safety ring: Polyester
  • Locking tag: Polyamid/stainless steel
  • Locking nut: Stainless steel in compliance with SS2346, ASTM® 303
  • Wetted materials: Stainless steel in compliance with 316L EN 1.4435, EN 10 272, ASME SA 479/ASTM® A 479

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
NOVASEPTUM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

NovaSeptum® GO sterile sampling unit with 15 mL single-use conical tube for small volume and endotoxin sampling. Closed design for aseptic process. Reduce product loss and risks of contamination. Available in single and manifold sampling format. Easily integrated into stainless-steel system via multi-use connection types (TC, in-line, ingold®).

NovaSeptum® GO sterile sampling unit with 15 mL single-use conical tube for small volume and endotoxin sampling. Closed design for aseptic process. Reduce product loss and risks of contamination. Available in single and manifold sampling format. Easily integrated into stainless-steel system via multi-use connection types (TC, in-line, ingold®).

NovaSeptum® GO sterile sampling unit with 15 mL single-use conical tube for small volume and endotoxin sampling. Closed design for aseptic process. Reduce product loss and risks of contamination. Available in single and manifold sampling format. Easily integrated into stainless-steel system via multi-use connection types (TC, in-line, ingold®).

NovaSeptum® GO sterile sampling unit with 15 mL single-use conical tube for small volume and endotoxin sampling. Closed design for aseptic process. Reduce product loss and risks of contamination. Available in single and manifold sampling format. Easily integrated into stainless-steel system via multi-use connection types (TC, in-line, ingold®).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service